Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23475108-23475379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NGDN
Entrez Gene ID
25983
Gene Name
neuroguidin
Gene Aliases
C14orf120, CANu1, LCP5, NGD, lpd-2
UniGene
Hs.9043
Gene Chromosome Location
Chr.14: 23469689-23478193 on Build GRCh38
Transcripts
NM_001042635.1 NM_015514.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
162900 c.238C>G p.L80V Chr.14: 23475264-23475264 on Build GRCh38
6333396 c.165G>T p.V55V Chr.14: 23475191-23475191 on Build GRCh38
5523276 c.171C>T p.D57D Chr.14: 23475197-23475197 on Build GRCh38
954795 c.228A>C p.S76S Chr.14: 23475254-23475254 on Build GRCh38
954793 c.223G>T p.A75S Chr.14: 23475249-23475249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147692634 Chr.14: 23475258-23475258 on Build GRCh38
rs771025811 Chr.14: 23475253-23475253 on Build GRCh38
rs777926351 Chr.14: 23475340-23475340 on Build GRCh38
rs898301133 Chr.14: 23475304-23475304 on Build GRCh38
rs781673767 Chr.14: 23475179-23475181 on Build GRCh38
rs575716360 Chr.14: 23475223-23475223 on Build GRCh38
rs556994556 Chr.14: 23475297-23475297 on Build GRCh38
rs753368438 Chr.14: 23475166-23475166 on Build GRCh38
rs2236260 Chr.14: 23475296-23475296 on Build GRCh38
rs747571214 Chr.14: 23475212-23475212 on Build GRCh38
rs767412590 Chr.14: 23475149-23475149 on Build GRCh38
rs749546420 Chr.14: 23475342-23475342 on Build GRCh38
rs990526186 Chr.14: 23475262-23475262 on Build GRCh38
rs768861698 Chr.14: 23475219-23475219 on Build GRCh38
rs370904707 Chr.14: 23475307-23475307 on Build GRCh38
rs781615592 Chr.14: 23475308-23475308 on Build GRCh38
rs757930257 Chr.14: 23475300-23475300 on Build GRCh38
rs951675655 Chr.14: 23475245-23475245 on Build GRCh38
rs761763010 Chr.14: 23475294-23475294 on Build GRCh38
rs368059261 Chr.14: 23475301-23475301 on Build GRCh38
rs773464767 Chr.14: 23475246-23475246 on Build GRCh38
rs763087979 Chr.14: 23475143-23475143 on Build GRCh38
rs559030516 Chr.14: 23475220-23475220 on Build GRCh38
rs756521586 Chr.14: 23475331-23475331 on Build GRCh38
rs778764448 Chr.14: 23475171-23475171 on Build GRCh38
rs756169174 Chr.14: 23475339-23475339 on Build GRCh38
rs958897463 Chr.14: 23475260-23475260 on Build GRCh38
rs567174488 Chr.14: 23475237-23475237 on Build GRCh38
rs1046937935 Chr.14: 23475317-23475317 on Build GRCh38
rs2236261 Chr.14: 23475305-23475305 on Build GRCh38
rs199643781 Chr.14: 23475247-23475247 on Build GRCh38
rs145265355 Chr.14: 23475177-23475177 on Build GRCh38
rs748483157 Chr.14: 23475309-23475309 on Build GRCh38
rs758021826 Chr.14: 23475200-23475200 on Build GRCh38
rs760342585 Chr.14: 23475158-23475158 on Build GRCh38
rs776470805 Chr.14: 23475275-23475275 on Build GRCh38
rs760668652 Chr.14: 23475274-23475274 on Build GRCh38
rs756980284 Chr.14: 23475168-23475168 on Build GRCh38
rs752751142 Chr.14: 23475303-23475304 on Build GRCh38
rs763983832 Chr.14: 23475163-23475163 on Build GRCh38
rs780402428 Chr.14: 23475211-23475211 on Build GRCh38
rs752733168 Chr.14: 23475154-23475154 on Build GRCh38

View Full Product Details