Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104275151-104275412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTO2
Entrez Gene ID
119391
Gene Name
glutathione S-transferase omega 2
Gene Aliases
GSTO 2-2, bA127L20.1
UniGene
Hs.107384
Gene Chromosome Location
Chr.10: 104267626-104304948 on Build GRCh38
Transcripts
NM_001191013.1 NM_001191014.1 NM_001191015.1 NM_183239.1 XM_006717624.3 XM_011539270.2 XM_011539271.1 XM_011539272.2 XM_011539273.2 XM_017015670.1 XM_017015671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6676627 c.103T>G p.S35A Chr.10: 104275294-104275294 on Build GRCh38
4914040 c.59C>A p.P20Q Chr.10: 104275250-104275250 on Build GRCh38
123506 c.59C>T p.P20L Chr.10: 104275250-104275250 on Build GRCh38
3782510 c.115C>A p.R39S Chr.10: 104275306-104275306 on Build GRCh38
5913042 c.124C>T p.L42F Chr.10: 104275315-104275315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771838005 Chr.10: 104275276-104275276 on Build GRCh38
rs747606911 Chr.10: 104275330-104275330 on Build GRCh38
rs138733888 Chr.10: 104275215-104275215 on Build GRCh38
rs774564678 Chr.10: 104275178-104275185 on Build GRCh38
rs371560878 Chr.10: 104275198-104275198 on Build GRCh38
rs376641881 Chr.10: 104275338-104275338 on Build GRCh38
rs753910980 Chr.10: 104275175-104275175 on Build GRCh38
rs151042116 Chr.10: 104275224-104275224 on Build GRCh38
rs766576465 Chr.10: 104275227-104275227 on Build GRCh38
rs45450498 Chr.10: 104275210-104275210 on Build GRCh38
rs754528243 Chr.10: 104275325-104275325 on Build GRCh38
rs35481786 Chr.10: 104275196-104275197 on Build GRCh38
rs752014949 Chr.10: 104275308-104275308 on Build GRCh38
rs200918851 Chr.10: 104275371-104275371 on Build GRCh38
rs773049534 Chr.10: 104275281-104275281 on Build GRCh38
rs748129163 Chr.10: 104275275-104275275 on Build GRCh38
rs386747184 Chr.10: 104275374-104275375 on Build GRCh38
rs201769633 Chr.10: 104275361-104275361 on Build GRCh38
rs769508351 Chr.10: 104275203-104275203 on Build GRCh38
rs766629755 Chr.10: 104275286-104275286 on Build GRCh38
rs995973190 Chr.10: 104275360-104275360 on Build GRCh38
rs755538006 Chr.10: 104275176-104275176 on Build GRCh38
rs750159791 Chr.10: 104275220-104275220 on Build GRCh38
rs778820913 Chr.10: 104275241-104275241 on Build GRCh38
rs941816362 Chr.10: 104275183-104275183 on Build GRCh38
rs140910092 Chr.10: 104275274-104275274 on Build GRCh38
rs776871728 Chr.10: 104275292-104275292 on Build GRCh38
rs773288830 Chr.10: 104275332-104275332 on Build GRCh38
rs34400162 Chr.10: 104275312-104275312 on Build GRCh38
rs879138026 Chr.10: 104275295-104275295 on Build GRCh38
rs202018124 Chr.10: 104275209-104275209 on Build GRCh38
rs748845877 Chr.10: 104275266-104275266 on Build GRCh38
rs748504034 Chr.10: 104275190-104275190 on Build GRCh38
rs910439079 Chr.10: 104275386-104275386 on Build GRCh38
rs941874457 Chr.10: 104275389-104275389 on Build GRCh38
rs199548115 Chr.10: 104275305-104275305 on Build GRCh38
rs370924472 Chr.10: 104275268-104275268 on Build GRCh38
rs956084993 Chr.10: 104275350-104275350 on Build GRCh38
rs753272891 Chr.10: 104275243-104275243 on Build GRCh38
rs992989656 Chr.10: 104275270-104275270 on Build GRCh38
rs760693254 Chr.10: 104275368-104275373 on Build GRCh38
rs762358128 Chr.10: 104275383-104275383 on Build GRCh38
rs747370101 Chr.10: 104275257-104275257 on Build GRCh38
rs755885559 Chr.10: 104275264-104275264 on Build GRCh38
rs765375682 Chr.10: 104275302-104275302 on Build GRCh38
rs752141268 Chr.10: 104275376-104275376 on Build GRCh38
rs745533473 Chr.10: 104275201-104275201 on Build GRCh38
rs762446344 Chr.10: 104275285-104275286 on Build GRCh38
rs866968480 Chr.10: 104275234-104275234 on Build GRCh38
rs762535564 Chr.10: 104275206-104275206 on Build GRCh38
rs779454821 Chr.10: 104275189-104275189 on Build GRCh38
rs536025328 Chr.10: 104275251-104275251 on Build GRCh38
rs778844897 Chr.10: 104275211-104275211 on Build GRCh38
rs759880709 Chr.10: 104275303-104275303 on Build GRCh38
rs112969459 Chr.10: 104275204-104275204 on Build GRCh38
rs1037466673 Chr.10: 104275186-104275186 on Build GRCh38
rs764681176 Chr.10: 104275306-104275306 on Build GRCh38
rs753968945 Chr.10: 104275229-104275229 on Build GRCh38
rs778489955 Chr.10: 104275326-104275326 on Build GRCh38
rs772520596 Chr.10: 104275192-104275192 on Build GRCh38
rs7923376 Chr.10: 104275374-104275374 on Build GRCh38
rs759812200 Chr.10: 104275373-104275373 on Build GRCh38
rs572727110 Chr.10: 104275311-104275311 on Build GRCh38
rs377152092 Chr.10: 104275265-104275265 on Build GRCh38
rs755021553 Chr.10: 104275234-104275234 on Build GRCh38
rs74154779 Chr.10: 104275375-104275375 on Build GRCh38
rs772498661 Chr.10: 104275316-104275330 on Build GRCh38
rs772096028 Chr.10: 104275346-104275346 on Build GRCh38
rs759631071 Chr.10: 104275298-104275298 on Build GRCh38
rs773104544 Chr.10: 104275357-104275357 on Build GRCh38
rs778208226 Chr.10: 104275256-104275256 on Build GRCh38

View Full Product Details