Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156245557-156245800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAQR6
Entrez Gene ID
79957
Gene Name
progestin and adipoQ receptor family member 6
Gene Aliases
PRdelta
UniGene
Hs.235873
Gene Chromosome Location
Chr.1: 156243321-156248488 on Build GRCh38
Transcripts
NM_001272104.1 NM_001272105.1 NM_001272106.1 NM_001272107.1 NM_001272108.1 NM_001272109.1 NM_001272110.1 NM_001272111.1 NM_001272112.1 NM_001272113.1 NM_024897.3 NM_198406.2 NR_073610.1 XM_005245494.3 XM_006711546.2 XM_006711547.3 XM_006711548.3 XM_006711552.2 XM_006711553.3 XM_011510000.2 XM_017002385.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7284398 c.479_481delCCT p.S160delS Chr.1: 156245566-156245568 on Build GRCh38
3677406 c.68-1G>C p.? Chr.1: 156245662-156245662 on Build GRCh38
6498711 c.75C>T p.A25A Chr.1: 156245654-156245654 on Build GRCh38
6359124 c.139G>C p.V47L Chr.1: 156245590-156245590 on Build GRCh38
897146 c.135C>A p.F45L Chr.1: 156245594-156245594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759436689 Chr.1: 156245691-156245691 on Build GRCh38
rs2540170 Chr.1: 156245755-156245755 on Build GRCh38
rs865916447 Chr.1: 156245711-156245711 on Build GRCh38
rs144076256 Chr.1: 156245594-156245594 on Build GRCh38
rs376734651 Chr.1: 156245779-156245779 on Build GRCh38
rs781386530 Chr.1: 156245756-156245756 on Build GRCh38
rs35270353 Chr.1: 156245640-156245640 on Build GRCh38
rs763971319 Chr.1: 156245737-156245737 on Build GRCh38
rs752722983 Chr.1: 156245708-156245708 on Build GRCh38
rs894414635 Chr.1: 156245772-156245772 on Build GRCh38
rs750142112 Chr.1: 156245590-156245590 on Build GRCh38
rs575679158 Chr.1: 156245621-156245621 on Build GRCh38
rs767215784 Chr.1: 156245700-156245700 on Build GRCh38
rs780098823 Chr.1: 156245642-156245642 on Build GRCh38
rs752560890 Chr.1: 156245625-156245625 on Build GRCh38
rs768827092 Chr.1: 156245623-156245623 on Build GRCh38
rs766138681 Chr.1: 156245678-156245678 on Build GRCh38
rs935332831 Chr.1: 156245599-156245599 on Build GRCh38
rs770451331 Chr.1: 156245769-156245769 on Build GRCh38
rs778930710 Chr.1: 156245634-156245634 on Build GRCh38
rs985307690 Chr.1: 156245612-156245612 on Build GRCh38
rs555819330 Chr.1: 156245657-156245657 on Build GRCh38
rs760437653 Chr.1: 156245773-156245774 on Build GRCh38
rs2075166 Chr.1: 156245771-156245771 on Build GRCh38
rs764836842 Chr.1: 156245587-156245587 on Build GRCh38
rs769479829 Chr.1: 156245665-156245665 on Build GRCh38
rs757391592 Chr.1: 156245628-156245628 on Build GRCh38
rs11552748 Chr.1: 156245648-156245648 on Build GRCh38
rs1018849732 Chr.1: 156245579-156245579 on Build GRCh38
rs748472010 Chr.1: 156245760-156245760 on Build GRCh38
rs772873870 Chr.1: 156245674-156245674 on Build GRCh38
rs748825501 Chr.1: 156245775-156245775 on Build GRCh38
rs375118362 Chr.1: 156245741-156245741 on Build GRCh38
rs914270330 Chr.1: 156245731-156245731 on Build GRCh38
rs372081403 Chr.1: 156245735-156245735 on Build GRCh38
rs747191688 Chr.1: 156245752-156245752 on Build GRCh38
rs770337685 Chr.1: 156245777-156245777 on Build GRCh38
rs374567613 Chr.1: 156245597-156245597 on Build GRCh38
rs766083677 Chr.1: 156245601-156245601 on Build GRCh38
rs773813500 Chr.1: 156245680-156245680 on Build GRCh38
rs544084502 Chr.1: 156245649-156245649 on Build GRCh38
rs1050066428 Chr.1: 156245704-156245704 on Build GRCh38
rs746136582 Chr.1: 156245635-156245635 on Build GRCh38
rs780380507 Chr.1: 156245745-156245745 on Build GRCh38
rs761445250 Chr.1: 156245585-156245585 on Build GRCh38
rs751406407 Chr.1: 156245613-156245613 on Build GRCh38
rs989470806 Chr.1: 156245611-156245611 on Build GRCh38
rs748637277 Chr.1: 156245663-156245663 on Build GRCh38
rs1044941433 Chr.1: 156245703-156245703 on Build GRCh38
rs774987605 Chr.1: 156245753-156245754 on Build GRCh38
rs923529123 Chr.1: 156245643-156245643 on Build GRCh38

View Full Product Details