Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34528777-34529026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PACSIN1
Entrez Gene ID
29993
Gene Name
protein kinase C and casein kinase substrate in neurons 1
Gene Aliases
SDPI
UniGene
Hs.520087
Gene Chromosome Location
Chr.6: 34466061-34536262 on Build GRCh38
Transcripts
NM_001199583.2 NM_020804.4 XM_011514541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1672321 c.422A>C p.K141T Chr.6: 34528843-34528843 on Build GRCh38
3625885 c.399G>A p.K133K Chr.6: 34528820-34528820 on Build GRCh38
6007211 c.364C>A p.H122N Chr.6: 34528785-34528785 on Build GRCh38
3625884 c.375C>T p.I125I Chr.6: 34528796-34528796 on Build GRCh38
5653651 c.454G>A p.E152K Chr.6: 34528875-34528875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146300537 Chr.6: 34528824-34528824 on Build GRCh38
rs373996594 Chr.6: 34528817-34528817 on Build GRCh38
rs771585407 Chr.6: 34528902-34528902 on Build GRCh38
rs370850796 Chr.6: 34528909-34528909 on Build GRCh38
rs1025402027 Chr.6: 34528960-34528960 on Build GRCh38
rs761691957 Chr.6: 34528918-34528918 on Build GRCh38
rs76107911 Chr.6: 34529003-34529003 on Build GRCh38
rs779167918 Chr.6: 34528894-34528894 on Build GRCh38
rs368944518 Chr.6: 34528811-34528811 on Build GRCh38
rs1054456847 Chr.6: 34528819-34528819 on Build GRCh38
rs751508946 Chr.6: 34528845-34528845 on Build GRCh38
rs755154125 Chr.6: 34528882-34528882 on Build GRCh38
rs926757377 Chr.6: 34529006-34529006 on Build GRCh38
rs558135547 Chr.6: 34528899-34528899 on Build GRCh38
rs562393363 Chr.6: 34528945-34528945 on Build GRCh38
rs540533288 Chr.6: 34528932-34528932 on Build GRCh38
rs932143177 Chr.6: 34528854-34528854 on Build GRCh38
rs1035854418 Chr.6: 34528934-34528934 on Build GRCh38
rs761745160 Chr.6: 34528861-34528861 on Build GRCh38
rs772099763 Chr.6: 34528919-34528919 on Build GRCh38
rs769598563 Chr.6: 34528816-34528816 on Build GRCh38
rs750404850 Chr.6: 34528924-34528924 on Build GRCh38
rs529590246 Chr.6: 34528974-34528974 on Build GRCh38
rs763568480 Chr.6: 34528839-34528839 on Build GRCh38
rs573209475 Chr.6: 34528913-34528913 on Build GRCh38
rs371006157 Chr.6: 34528874-34528874 on Build GRCh38
rs749252825 Chr.6: 34528902-34528903 on Build GRCh38
rs758493750 Chr.6: 34528898-34528898 on Build GRCh38
rs1009533336 Chr.6: 34528956-34528956 on Build GRCh38
rs746200947 Chr.6: 34528903-34528903 on Build GRCh38
rs750241333 Chr.6: 34528881-34528881 on Build GRCh38
rs752914821 Chr.6: 34528895-34528895 on Build GRCh38
rs770104515 Chr.6: 34528908-34528908 on Build GRCh38
rs773748210 Chr.6: 34528917-34528917 on Build GRCh38
rs1027319890 Chr.6: 34528975-34528975 on Build GRCh38
rs775044885 Chr.6: 34528823-34528823 on Build GRCh38
rs768144014 Chr.6: 34528915-34528915 on Build GRCh38

View Full Product Details