Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75728869-75729094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOGAT2
Entrez Gene ID
80168
Gene Name
monoacylglycerol O-acyltransferase 2
Gene Aliases
DGAT2L5, DGAT2L5., MGAT2, hDC5
UniGene
Hs.288568
Gene Chromosome Location
Chr.11: 75717819-75732958 on Build GRCh38
Transcripts
NM_025098.3 XM_011545267.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2154963 c.736A>T p.I246F Chr.11: 75728875-75728875 on Build GRCh38
4702834 c.804G>A p.Q268Q Chr.11: 75728943-75728943 on Build GRCh38
2154965 c.737T>G p.I246S Chr.11: 75728876-75728876 on Build GRCh38
4036756 c.745C>T p.R249W Chr.11: 75728884-75728884 on Build GRCh38
4757643 c.811T>G p.F271V Chr.11: 75728950-75728950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776178184 Chr.11: 75728955-75728955 on Build GRCh38
rs541032350 Chr.11: 75728999-75728999 on Build GRCh38
rs752336428 Chr.11: 75729006-75729006 on Build GRCh38
rs760565995 Chr.11: 75728977-75728977 on Build GRCh38
rs771259921 Chr.11: 75728945-75728945 on Build GRCh38
rs369300429 Chr.11: 75729029-75729029 on Build GRCh38
rs550221599 Chr.11: 75729043-75729043 on Build GRCh38
rs375395725 Chr.11: 75729022-75729022 on Build GRCh38
rs551366609 Chr.11: 75729044-75729044 on Build GRCh38
rs374126807 Chr.11: 75728898-75728898 on Build GRCh38
rs368435356 Chr.11: 75728922-75728922 on Build GRCh38
rs562033079 Chr.11: 75728907-75728907 on Build GRCh38
rs779490736 Chr.11: 75728911-75728911 on Build GRCh38
rs747820873 Chr.11: 75728940-75728940 on Build GRCh38
rs12281506 Chr.11: 75729025-75729025 on Build GRCh38
rs1036949202 Chr.11: 75729059-75729059 on Build GRCh38
rs867093633 Chr.11: 75729049-75729049 on Build GRCh38
rs770451237 Chr.11: 75728954-75728954 on Build GRCh38
rs768718260 Chr.11: 75729041-75729041 on Build GRCh38
rs766273873 Chr.11: 75728983-75728983 on Build GRCh38
rs377311369 Chr.11: 75728968-75728968 on Build GRCh38
rs758008390 Chr.11: 75729017-75729017 on Build GRCh38
rs370837360 Chr.11: 75728969-75728969 on Build GRCh38
rs770328531 Chr.11: 75729028-75729028 on Build GRCh38
rs768917564 Chr.11: 75728965-75728965 on Build GRCh38
rs754525542 Chr.11: 75728929-75728929 on Build GRCh38
rs763099534 Chr.11: 75728962-75728962 on Build GRCh38
rs778612784 Chr.11: 75728930-75728930 on Build GRCh38
rs761657918 Chr.11: 75728892-75728892 on Build GRCh38
rs959390660 Chr.11: 75728897-75728897 on Build GRCh38
rs767289035 Chr.11: 75728971-75728971 on Build GRCh38
rs750312025 Chr.11: 75728902-75728902 on Build GRCh38
rs750208208 Chr.11: 75728972-75728972 on Build GRCh38
rs781423297 Chr.11: 75728946-75728946 on Build GRCh38
rs868819849 Chr.11: 75729020-75729020 on Build GRCh38
rs749880145 Chr.11: 75729030-75729030 on Build GRCh38
rs772405933 Chr.11: 75728942-75728942 on Build GRCh38
rs746332568 Chr.11: 75728949-75728949 on Build GRCh38
rs778289450 Chr.11: 75729004-75729004 on Build GRCh38

View Full Product Details