Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24915538-24915809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEFM
Entrez Gene ID
4741
Gene Name
neurofilament, medium polypeptide
Gene Aliases
NEF3, NF-M, NFM
UniGene
Hs.458657
Gene Chromosome Location
Chr.8: 24913761-24919093 on Build GRCh38
Transcripts
NM_001105541.1 NM_005382.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
402978 c.1168G>T p.V390F Chr.8: 24915692-24915692 on Build GRCh38
262380 c.1111C>T p.R371W Chr.8: 24915635-24915635 on Build GRCh38
1098386 c.1194C>T p.I398I Chr.8: 24915718-24915718 on Build GRCh38
7379297 c.1130T>A p.M377K Chr.8: 24915654-24915654 on Build GRCh38
1755672 c.1144C>T p.R382C Chr.8: 24915668-24915668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759465720 Chr.8: 24915660-24915660 on Build GRCh38
rs200024394 Chr.8: 24915672-24915672 on Build GRCh38
rs757939700 Chr.8: 24915582-24915582 on Build GRCh38
rs370943444 Chr.8: 24915605-24915605 on Build GRCh38
rs776101701 Chr.8: 24915768-24915768 on Build GRCh38
rs753109080 Chr.8: 24915558-24915558 on Build GRCh38
rs747541164 Chr.8: 24915772-24915772 on Build GRCh38
rs775844692 Chr.8: 24915668-24915668 on Build GRCh38
rs149916684 Chr.8: 24915787-24915787 on Build GRCh38
rs781676915 Chr.8: 24915734-24915734 on Build GRCh38
rs59419000 Chr.8: 24915628-24915628 on Build GRCh38
rs988236952 Chr.8: 24915573-24915573 on Build GRCh38
rs774502591 Chr.8: 24915656-24915656 on Build GRCh38
rs762619609 Chr.8: 24915687-24915687 on Build GRCh38
rs764924154 Chr.8: 24915569-24915569 on Build GRCh38
rs887499286 Chr.8: 24915663-24915663 on Build GRCh38
rs757503617 Chr.8: 24915757-24915757 on Build GRCh38
rs780927497 Chr.8: 24915608-24915608 on Build GRCh38
rs111844007 Chr.8: 24915665-24915665 on Build GRCh38
rs771228026 Chr.8: 24915643-24915643 on Build GRCh38
rs764406988 Chr.8: 24915670-24915670 on Build GRCh38
rs73545170 Chr.8: 24915778-24915778 on Build GRCh38
rs769106395 Chr.8: 24915774-24915774 on Build GRCh38
rs781549347 Chr.8: 24915637-24915637 on Build GRCh38
rs781249697 Chr.8: 24915743-24915743 on Build GRCh38
rs779649709 Chr.8: 24915583-24915583 on Build GRCh38
rs757327763 Chr.8: 24915735-24915735 on Build GRCh38
rs766276484 Chr.8: 24915739-24915739 on Build GRCh38
rs141854965 Chr.8: 24915709-24915709 on Build GRCh38
rs186811095 Chr.8: 24915746-24915746 on Build GRCh38
rs778996520 Chr.8: 24915760-24915760 on Build GRCh38
rs750987798 Chr.8: 24915595-24915595 on Build GRCh38
rs749490843 Chr.8: 24915638-24915638 on Build GRCh38
rs746015124 Chr.8: 24915764-24915764 on Build GRCh38
rs201942700 Chr.8: 24915752-24915752 on Build GRCh38
rs200056948 Chr.8: 24915691-24915691 on Build GRCh38
rs905190963 Chr.8: 24915729-24915729 on Build GRCh38
rs565997828 Chr.8: 24915756-24915756 on Build GRCh38
rs753836154 Chr.8: 24915756-24915756 on Build GRCh38
rs761072893 Chr.8: 24915669-24915669 on Build GRCh38
rs750051140 Chr.8: 24915579-24915579 on Build GRCh38
rs575280559 Chr.8: 24915786-24915786 on Build GRCh38
rs377590256 Chr.8: 24915776-24915776 on Build GRCh38
rs748043489 Chr.8: 24915609-24915609 on Build GRCh38
rs751655016 Chr.8: 24915587-24915588 on Build GRCh38
rs534915013 Chr.8: 24915711-24915711 on Build GRCh38
rs769700786 Chr.8: 24915610-24915610 on Build GRCh38
rs577724259 Chr.8: 24915767-24915767 on Build GRCh38
rs754833080 Chr.8: 24915773-24915787 on Build GRCh38

View Full Product Details