Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50099406-50099645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOV10L1
Entrez Gene ID
54456
Gene Name
Mov10 RISC complex RNA helicase like 1
Gene Aliases
CHAMP, DJ402G11.8
UniGene
Hs.62880
Gene Chromosome Location
Chr.22: 50090006-50161705 on Build GRCh38
Transcripts
NM_001164104.1 NM_001164105.1 NM_018995.2 XM_005261923.3 XM_011530696.1 XM_011530697.1 XM_011530698.1 XM_011530699.1 XM_011530700.2 XM_011530701.1 XM_011530702.1 XM_011530704.1 XM_017028833.1 XM_017028834.1 XM_017028837.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1714518 c.379G>A p.E127K Chr.22: 50099599-50099599 on Build GRCh38
1714517 c.439G>A p.E147K Chr.22: 50099599-50099599 on Build GRCh38
4983731 c.252C>T p.D84D Chr.22: 50099472-50099472 on Build GRCh38
4485224 c.230C>T p.A77V Chr.22: 50099450-50099450 on Build GRCh38
6714503 c.331G>A p.A111T Chr.22: 50099551-50099551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190821816 Chr.22: 50099532-50099532 on Build GRCh38
rs770996969 Chr.22: 50099503-50099503 on Build GRCh38
rs781497595 Chr.22: 50099582-50099582 on Build GRCh38
rs775255128 Chr.22: 50099513-50099513 on Build GRCh38
rs201595098 Chr.22: 50099487-50099487 on Build GRCh38
rs763939565 Chr.22: 50099544-50099544 on Build GRCh38
rs149582724 Chr.22: 50099510-50099510 on Build GRCh38
rs779422667 Chr.22: 50099614-50099614 on Build GRCh38
rs373495128 Chr.22: 50099462-50099462 on Build GRCh38
rs140532446 Chr.22: 50099551-50099551 on Build GRCh38
rs765661527 Chr.22: 50099475-50099475 on Build GRCh38
rs12169319 Chr.22: 50099472-50099472 on Build GRCh38
rs762299641 Chr.22: 50099473-50099473 on Build GRCh38
rs760228272 Chr.22: 50099522-50099522 on Build GRCh38
rs150821412 Chr.22: 50099459-50099459 on Build GRCh38
rs757077590 Chr.22: 50099598-50099598 on Build GRCh38
rs767221349 Chr.22: 50099456-50099456 on Build GRCh38
rs753707707 Chr.22: 50099592-50099592 on Build GRCh38
rs758912625 Chr.22: 50099446-50099446 on Build GRCh38
rs146380250 Chr.22: 50099599-50099599 on Build GRCh38
rs200845485 Chr.22: 50099528-50099528 on Build GRCh38
rs777698774 Chr.22: 50099488-50099488 on Build GRCh38
rs780357815 Chr.22: 50099484-50099484 on Build GRCh38
rs369455937 Chr.22: 50099495-50099495 on Build GRCh38
rs762634864 Chr.22: 50099490-50099490 on Build GRCh38
rs750519145 Chr.22: 50099480-50099480 on Build GRCh38
rs751867186 Chr.22: 50099556-50099556 on Build GRCh38
rs763137022 Chr.22: 50099550-50099550 on Build GRCh38
rs143809250 Chr.22: 50099577-50099577 on Build GRCh38
rs773994274 Chr.22: 50099434-50099434 on Build GRCh38
rs774963444 Chr.22: 50099461-50099461 on Build GRCh38
rs758364053 Chr.22: 50099483-50099483 on Build GRCh38
rs778944666 Chr.22: 50099504-50099504 on Build GRCh38
rs764341057 Chr.22: 50099469-50099469 on Build GRCh38
rs376465916 Chr.22: 50099482-50099482 on Build GRCh38
rs746458711 Chr.22: 50099615-50099615 on Build GRCh38
rs185518372 Chr.22: 50099519-50099519 on Build GRCh38
rs867320608 Chr.22: 50099494-50099494 on Build GRCh38
rs146098008 Chr.22: 50099538-50099538 on Build GRCh38
rs745767983 Chr.22: 50099603-50099603 on Build GRCh38
rs745413330 Chr.22: 50099506-50099506 on Build GRCh38
rs771668569 Chr.22: 50099605-50099605 on Build GRCh38
rs369649531 Chr.22: 50099430-50099430 on Build GRCh38
rs751702200 Chr.22: 50099486-50099486 on Build GRCh38
rs759930568 Chr.22: 50099460-50099460 on Build GRCh38
rs771451406 Chr.22: 50099508-50099508 on Build GRCh38

View Full Product Details