Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163463651-163463920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMMR
Entrez Gene ID
3161
Gene Name
hyaluronan mediated motility receptor
Gene Aliases
CD168, IHABP, RHAMM
UniGene
Hs.740467
Gene Chromosome Location
Chr.5: 163460511-163491945 on Build GRCh38
Transcripts
NM_001142556.1 NM_001142557.1 NM_012484.2 NM_012485.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4599713 c.82A>G p.T28A Chr.5: 163463891-163463891 on Build GRCh38
6470153 c.76G>T p.V26F Chr.5: 163463885-163463885 on Build GRCh38
6328425 c.88G>T p.E30* Chr.5: 163463897-163463897 on Build GRCh38
3248020 c.104C>T p.P35L Chr.5: 163463913-163463913 on Build GRCh38
6328426 c.111C>T p.S37S Chr.5: 163463920-163463920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970850927 Chr.5: 163463735-163463735 on Build GRCh38
rs752901172 Chr.5: 163463886-163463886 on Build GRCh38
rs973648235 Chr.5: 163463681-163463681 on Build GRCh38
rs936059448 Chr.5: 163463810-163463810 on Build GRCh38
rs543982329 Chr.5: 163463857-163463857 on Build GRCh38
rs553750421 Chr.5: 163463874-163463874 on Build GRCh38
rs926037028 Chr.5: 163463798-163463798 on Build GRCh38
rs963126012 Chr.5: 163463679-163463679 on Build GRCh38
rs751533811 Chr.5: 163463863-163463863 on Build GRCh38
rs530497637 Chr.5: 163463693-163463693 on Build GRCh38
rs768549675 Chr.5: 163463832-163463832 on Build GRCh38
rs767696177 Chr.5: 163463875-163463875 on Build GRCh38
rs946067701 Chr.5: 163463724-163463724 on Build GRCh38
rs551901312 Chr.5: 163463705-163463705 on Build GRCh38
rs570460506 Chr.5: 163463743-163463743 on Build GRCh38
rs775205873 Chr.5: 163463811-163463811 on Build GRCh38
rs759490251 Chr.5: 163463698-163463698 on Build GRCh38
rs374802971 Chr.5: 163463830-163463830 on Build GRCh38
rs992731467 Chr.5: 163463699-163463699 on Build GRCh38
rs765374948 Chr.5: 163463886-163463886 on Build GRCh38
rs1041669572 Chr.5: 163463793-163463793 on Build GRCh38
rs766502267 Chr.5: 163463862-163463862 on Build GRCh38
rs917145451 Chr.5: 163463711-163463711 on Build GRCh38
rs1051367208 Chr.5: 163463849-163463849 on Build GRCh38
rs916764564 Chr.5: 163463690-163463690 on Build GRCh38
rs535313154 Chr.5: 163463834-163463834 on Build GRCh38

View Full Product Details