Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 123025767-123026002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIC
Entrez Gene ID
5480
Gene Name
peptidylprolyl isomerase C
Gene Aliases
CYPC
UniGene
Hs.110364
Gene Chromosome Location
Chr.5: 123023383-123036730 on Build GRCh38
Transcripts
NM_000943.4 XM_017009395.1

Gene Symbol
SNX24
Entrez Gene ID
28966
Gene Name
sorting nexin 24
Gene Aliases
PRO1284, SBBI31
UniGene
Hs.483200
Gene Chromosome Location
Chr.5: 122845465-123029318 on Build GRCh38
Transcripts
NM_000943.4 XM_017009395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6560526 c.347C>A p.T116K Chr.5: 123025947-123025947 on Build GRCh38
3135149 c.410C>G p.A137G Chr.5: 123025884-123025884 on Build GRCh38
1319294 c.472G>A p.G158S Chr.5: 123025822-123025822 on Build GRCh38
72262 c.466T>G p.L156V Chr.5: 123025828-123025828 on Build GRCh38
5402031 c.369G>T p.K123N Chr.5: 123025925-123025925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141769515 Chr.5: 123025905-123025905 on Build GRCh38
rs774020150 Chr.5: 123025953-123025953 on Build GRCh38
rs781284487 Chr.5: 123025908-123025908 on Build GRCh38
rs755709937 Chr.5: 123025885-123025885 on Build GRCh38
rs200755681 Chr.5: 123025917-123025917 on Build GRCh38
rs775851495 Chr.5: 123025914-123025914 on Build GRCh38
rs74557853 Chr.5: 123025886-123025886 on Build GRCh38
rs766984428 Chr.5: 123025869-123025869 on Build GRCh38
rs763860453 Chr.5: 123025834-123025834 on Build GRCh38
rs768438238 Chr.5: 123025931-123025931 on Build GRCh38
rs747487256 Chr.5: 123025901-123025901 on Build GRCh38
rs761668325 Chr.5: 123025856-123025856 on Build GRCh38
rs747531352 Chr.5: 123025811-123025811 on Build GRCh38
rs1051358562 Chr.5: 123025898-123025898 on Build GRCh38
rs45560036 Chr.5: 123025815-123025815 on Build GRCh38
rs370628209 Chr.5: 123025910-123025910 on Build GRCh38
rs751782851 Chr.5: 123025808-123025808 on Build GRCh38
rs775904353 Chr.5: 123025829-123025829 on Build GRCh38
rs753525453 Chr.5: 123025888-123025888 on Build GRCh38
rs921257993 Chr.5: 123025836-123025836 on Build GRCh38
rs986807895 Chr.5: 123025906-123025906 on Build GRCh38
rs758807912 Chr.5: 123025808-123025808 on Build GRCh38
rs778342861 Chr.5: 123025810-123025810 on Build GRCh38
rs762124000 Chr.5: 123025814-123025815 on Build GRCh38
rs574236685 Chr.5: 123025893-123025893 on Build GRCh38
rs770035332 Chr.5: 123025911-123025911 on Build GRCh38
rs945520075 Chr.5: 123025947-123025947 on Build GRCh38
rs143084462 Chr.5: 123025822-123025822 on Build GRCh38
rs758752556 Chr.5: 123025889-123025889 on Build GRCh38
rs368146054 Chr.5: 123025823-123025823 on Build GRCh38
rs770988129 Chr.5: 123025812-123025812 on Build GRCh38
rs761768432 Chr.5: 123025974-123025974 on Build GRCh38
rs556627573 Chr.5: 123025832-123025832 on Build GRCh38
rs190810674 Chr.5: 123025842-123025842 on Build GRCh38

View Full Product Details