Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161229484-161229757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR1I3
Entrez Gene ID
9970
Gene Name
nuclear receptor subfamily 1 group I member 3
Gene Aliases
CAR, CAR1, MB67
UniGene
Hs.349642
Gene Chromosome Location
Chr.1: 161229666-161238623 on Build GRCh38
Transcripts
NM_001077469.2 NM_001077470.2 NM_001077471.2 NM_001077472.2 NM_001077473.2 NM_001077474.2 NM_001077475.2 NM_001077476.2 NM_001077477.2 NM_001077478.2 NM_001077479.2 NM_001077480.2 NM_001077481.2 NM_001077482.2 NM_005122.4 XM_005245693.4 XM_005245694.4 XM_005245697.4 XM_011510237.2 NM_001286373.1 NM_001286374.1 NM_032174.5 XM_006711572.2 XM_011510057.2 XM_017002481.1 XM_017002482.1

Gene Symbol
TOMM40L
Entrez Gene ID
84134
Gene Name
translocase of outer mitochondrial membrane 40 like
Gene Aliases
TOMM40B
UniGene
Hs.321653
Gene Chromosome Location
Chr.1: 161225939-161230746 on Build GRCh38
Transcripts
NM_001077469.2 NM_001077470.2 NM_001077471.2 NM_001077472.2 NM_001077473.2 NM_001077474.2 NM_001077475.2 NM_001077476.2 NM_001077477.2 NM_001077478.2 NM_001077479.2 NM_001077480.2 NM_001077481.2 NM_001077482.2 NM_005122.4 XM_005245693.4 XM_005245694.4 XM_005245697.4 XM_011510237.2 NM_001286373.1 NM_001286374.1 NM_032174.5 XM_006711572.2 XM_011510057.2 XM_017002481.1 XM_017002482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4519135 c.993_994GG>AA p.G332S Chr.1: 161229711-161229712 on Build GRCh38
4563121 c.959G>A p.W320* Chr.1: 161229746-161229746 on Build GRCh38
898540 c.991C>T p.P331S Chr.1: 161229714-161229714 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs899072992 Chr.1: 161229536-161229536 on Build GRCh38
rs34645321 Chr.1: 161229712-161229712 on Build GRCh38
rs764887689 Chr.1: 161229559-161229559 on Build GRCh38
rs769025799 Chr.1: 161229645-161229645 on Build GRCh38
rs778081793 Chr.1: 161229704-161229704 on Build GRCh38
rs377309702 Chr.1: 161229638-161229638 on Build GRCh38
rs569420131 Chr.1: 161229565-161229565 on Build GRCh38
rs776956399 Chr.1: 161229650-161229650 on Build GRCh38
rs3813629 Chr.1: 161229728-161229728 on Build GRCh38
rs368521829 Chr.1: 161229674-161229674 on Build GRCh38
rs773748252 Chr.1: 161229690-161229690 on Build GRCh38
rs746343817 Chr.1: 161229635-161229635 on Build GRCh38
rs201243903 Chr.1: 161229713-161229713 on Build GRCh38
rs35937139 Chr.1: 161229599-161229599 on Build GRCh38
rs1008954747 Chr.1: 161229569-161229569 on Build GRCh38
rs927862863 Chr.1: 161229618-161229618 on Build GRCh38
rs747050280 Chr.1: 161229642-161229642 on Build GRCh38
rs971545687 Chr.1: 161229684-161229684 on Build GRCh38
rs755279262 Chr.1: 161229733-161229733 on Build GRCh38
rs752531073 Chr.1: 161229650-161229650 on Build GRCh38
rs2307417 Chr.1: 161229640-161229640 on Build GRCh38
rs903309268 Chr.1: 161229619-161229619 on Build GRCh38
rs748001601 Chr.1: 161229651-161229651 on Build GRCh38
rs962838434 Chr.1: 161229552-161229552 on Build GRCh38
rs752324753 Chr.1: 161229705-161229705 on Build GRCh38
rs1037810089 Chr.1: 161229511-161229511 on Build GRCh38
rs981786446 Chr.1: 161229685-161229685 on Build GRCh38
rs766275202 Chr.1: 161229683-161229683 on Build GRCh38
rs35520307 Chr.1: 161229641-161229641 on Build GRCh38
rs767195966 Chr.1: 161229697-161229697 on Build GRCh38
rs940468355 Chr.1: 161229710-161229710 on Build GRCh38
rs769694595 Chr.1: 161229652-161229652 on Build GRCh38
rs758152799 Chr.1: 161229608-161229608 on Build GRCh38
rs537248708 Chr.1: 161229726-161229726 on Build GRCh38
rs939195803 Chr.1: 161229625-161229625 on Build GRCh38
rs77217925 Chr.1: 161229665-161229665 on Build GRCh38
rs780564353 Chr.1: 161229734-161229734 on Build GRCh38
rs1050249185 Chr.1: 161229563-161229563 on Build GRCh38
rs749380602 Chr.1: 161229731-161229731 on Build GRCh38
rs756461027 Chr.1: 161229651-161229652 on Build GRCh38
rs958967385 Chr.1: 161229654-161229654 on Build GRCh38
rs759006086 Chr.1: 161229692-161229692 on Build GRCh38
rs1047710118 Chr.1: 161229614-161229614 on Build GRCh38
rs2307416 Chr.1: 161229711-161229711 on Build GRCh38

View Full Product Details