Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 77328897-77329152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE8B
Entrez Gene ID
8622
Gene Name
phosphodiesterase 8B
Gene Aliases
ADSD, PPNAD3
UniGene
Hs.584830
Gene Chromosome Location
Chr.5: 77180252-77428256 on Build GRCh38
Transcripts
NM_001029851.2 NM_001029852.2 NM_001029853.2 NM_001029854.2 NM_003719.3 XM_005248621.4 XM_005248623.4 XM_005248624.4 XM_006714725.3 XM_006714726.3 XM_011543699.2 XM_011543700.2 XM_011543701.2 XM_011543702.2 XM_011543703.2 XM_011543704.2 XM_017010003.1 XM_017010004.1 XM_017010005.1 XM_017010006.1 XM_017010007.1 XM_017010008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4502331 c.610C>A p.P204T Chr.5: 77329017-77329017 on Build GRCh38
5403850 c.610C>T p.P204S Chr.5: 77329017-77329017 on Build GRCh38
6286990 c.650+5G>T p.? Chr.5: 77329062-77329062 on Build GRCh38
5439001 c.623C>T p.T208M Chr.5: 77329030-77329030 on Build GRCh38
3941376 c.635C>T p.A212V Chr.5: 77329042-77329042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768184312 Chr.5: 77328959-77328959 on Build GRCh38
rs142671242 Chr.5: 77329052-77329052 on Build GRCh38
rs370337970 Chr.5: 77328981-77328981 on Build GRCh38
rs986929584 Chr.5: 77329087-77329087 on Build GRCh38
rs749085656 Chr.5: 77329109-77329109 on Build GRCh38
rs745826346 Chr.5: 77329014-77329014 on Build GRCh38
rs58410936 Chr.5: 77329094-77329094 on Build GRCh38
rs769683936 Chr.5: 77329099-77329099 on Build GRCh38
rs540259869 Chr.5: 77328941-77328941 on Build GRCh38
rs370696702 Chr.5: 77329022-77329022 on Build GRCh38
rs567244221 Chr.5: 77329122-77329122 on Build GRCh38
rs756459332 Chr.5: 77328978-77328978 on Build GRCh38
rs751181028 Chr.5: 77328953-77328953 on Build GRCh38
rs767163990 Chr.5: 77329043-77329043 on Build GRCh38
rs1017163091 Chr.5: 77329005-77329005 on Build GRCh38
rs754217682 Chr.5: 77329054-77329054 on Build GRCh38
rs766765735 Chr.5: 77329053-77329053 on Build GRCh38
rs865794141 Chr.5: 77328935-77328935 on Build GRCh38
rs763592439 Chr.5: 77328948-77328948 on Build GRCh38
rs776512625 Chr.5: 77329006-77329006 on Build GRCh38
rs200516880 Chr.5: 77328991-77328991 on Build GRCh38
rs138088764 Chr.5: 77329106-77329106 on Build GRCh38
rs375151381 Chr.5: 77328970-77328970 on Build GRCh38
rs368417608 Chr.5: 77329091-77329091 on Build GRCh38
rs373692136 Chr.5: 77328960-77328960 on Build GRCh38
rs137986191 Chr.5: 77329031-77329031 on Build GRCh38
rs779147669 Chr.5: 77329074-77329074 on Build GRCh38
rs755226259 Chr.5: 77328989-77328989 on Build GRCh38
rs755205148 Chr.5: 77329065-77329065 on Build GRCh38
rs761444971 Chr.5: 77329041-77329041 on Build GRCh38
rs770825592 Chr.5: 77329001-77329001 on Build GRCh38
rs377536938 Chr.5: 77329000-77329000 on Build GRCh38
rs748403580 Chr.5: 77329078-77329078 on Build GRCh38
rs758426659 Chr.5: 77329086-77329086 on Build GRCh38
rs757821773 Chr.5: 77328958-77328958 on Build GRCh38
rs749463385 Chr.5: 77328983-77328983 on Build GRCh38
rs372062422 Chr.5: 77329070-77329070 on Build GRCh38
rs750904456 Chr.5: 77329051-77329051 on Build GRCh38

View Full Product Details