Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38881538-38881797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSMR
Entrez Gene ID
9180
Gene Name
oncostatin M receptor
Gene Aliases
IL-31R-beta, IL-31RB, OSMRB, PLCA1
UniGene
Hs.120658
Gene Chromosome Location
Chr.5: 38845858-38945579 on Build GRCh38
Transcripts
NM_001168355.2 NM_001323504.1 NM_001323505.1 NM_001323506.1 NM_001323507.1 NM_003999.2 XM_005248384.1 XM_005248386.2 XM_005248387.2 XM_011514161.2 XM_017010019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5164614 c.357C>T p.D119D Chr.5: 38881703-38881703 on Build GRCh38
5099838 c.302C>A p.S101Y Chr.5: 38881648-38881648 on Build GRCh38
5545522 c.290G>A p.W97* Chr.5: 38881636-38881636 on Build GRCh38
737823 c.309C>T p.L103L Chr.5: 38881655-38881655 on Build GRCh38
737822 c.358G>A p.D120N Chr.5: 38881704-38881704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918754155 Chr.5: 38881676-38881676 on Build GRCh38
rs759903125 Chr.5: 38881658-38881658 on Build GRCh38
rs762120200 Chr.5: 38881639-38881639 on Build GRCh38
rs185498494 Chr.5: 38881679-38881679 on Build GRCh38
rs751337172 Chr.5: 38881703-38881703 on Build GRCh38
rs889924104 Chr.5: 38881748-38881748 on Build GRCh38
rs768776188 Chr.5: 38881629-38881629 on Build GRCh38
rs773759068 Chr.5: 38881655-38881655 on Build GRCh38
rs757178327 Chr.5: 38881569-38881569 on Build GRCh38
rs375677074 Chr.5: 38881571-38881571 on Build GRCh38
rs774731604 Chr.5: 38881635-38881635 on Build GRCh38
rs866828886 Chr.5: 38881613-38881613 on Build GRCh38
rs768020747 Chr.5: 38881675-38881675 on Build GRCh38
rs927108711 Chr.5: 38881568-38881568 on Build GRCh38
rs370592814 Chr.5: 38881704-38881704 on Build GRCh38
rs758018785 Chr.5: 38881689-38881689 on Build GRCh38
rs772643747 Chr.5: 38881772-38881774 on Build GRCh38
rs780223229 Chr.5: 38881756-38881756 on Build GRCh38
rs990890871 Chr.5: 38881573-38881573 on Build GRCh38
rs1007071945 Chr.5: 38881760-38881760 on Build GRCh38
rs764668031 Chr.5: 38881768-38881769 on Build GRCh38
rs768885996 Chr.5: 38881769-38881769 on Build GRCh38
rs149778657 Chr.5: 38881738-38881738 on Build GRCh38
rs776215037 Chr.5: 38881764-38881765 on Build GRCh38
rs749351551 Chr.5: 38881765-38881765 on Build GRCh38
rs963105065 Chr.5: 38881622-38881622 on Build GRCh38
rs752210340 Chr.5: 38881680-38881680 on Build GRCh38
rs745864607 Chr.5: 38881736-38881736 on Build GRCh38
rs866378619 Chr.5: 38881603-38881603 on Build GRCh38
rs749451969 Chr.5: 38881624-38881624 on Build GRCh38
rs768041549 Chr.5: 38881640-38881640 on Build GRCh38
rs763683909 Chr.5: 38881699-38881699 on Build GRCh38
rs770052542 Chr.5: 38881594-38881594 on Build GRCh38
rs761514897 Chr.5: 38881765-38881766 on Build GRCh38
rs775847490 Chr.5: 38881601-38881601 on Build GRCh38

View Full Product Details