Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69699099-69699368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF19
Entrez Gene ID
9965
Gene Name
fibroblast growth factor 19
Gene Aliases
-
UniGene
Hs.249200
Gene Chromosome Location
Chr.11: 69698238-69704338 on Build GRCh38
Transcripts
NM_005117.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6924761 c.592G>C p.D198H Chr.11: 69699321-69699321 on Build GRCh38
6949737 c.571C>T p.P191S Chr.11: 69699342-69699342 on Build GRCh38
3870051 c.566C>T p.S189F Chr.11: 69699347-69699347 on Build GRCh38
6928579 c.614G>C p.G205A Chr.11: 69699299-69699299 on Build GRCh38
6955729 c.574C>A p.L192M Chr.11: 69699339-69699339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138132534 Chr.11: 69699288-69699288 on Build GRCh38
rs747731982 Chr.11: 69699273-69699273 on Build GRCh38
rs746692820 Chr.11: 69699297-69699297 on Build GRCh38
rs185560971 Chr.11: 69699201-69699201 on Build GRCh38
rs756366989 Chr.11: 69699243-69699243 on Build GRCh38
rs750468710 Chr.11: 69699333-69699333 on Build GRCh38
rs749561746 Chr.11: 69699251-69699251 on Build GRCh38
rs150092110 Chr.11: 69699343-69699343 on Build GRCh38
rs570500898 Chr.11: 69699193-69699193 on Build GRCh38
rs769260055 Chr.11: 69699255-69699255 on Build GRCh38
rs775268479 Chr.11: 69699326-69699326 on Build GRCh38
rs779279247 Chr.11: 69699266-69699266 on Build GRCh38
rs948787955 Chr.11: 69699187-69699187 on Build GRCh38
rs898223642 Chr.11: 69699289-69699289 on Build GRCh38
rs750699805 Chr.11: 69699227-69699227 on Build GRCh38
rs957399787 Chr.11: 69699304-69699304 on Build GRCh38
rs1055438741 Chr.11: 69699198-69699198 on Build GRCh38
rs1019112835 Chr.11: 69699250-69699250 on Build GRCh38
rs766637971 Chr.11: 69699311-69699311 on Build GRCh38
rs781615836 Chr.11: 69699221-69699221 on Build GRCh38
rs771545242 Chr.11: 69699278-69699278 on Build GRCh38
rs914632483 Chr.11: 69699168-69699168 on Build GRCh38
rs762938267 Chr.11: 69699330-69699330 on Build GRCh38
rs73524727 Chr.11: 69699195-69699195 on Build GRCh38
rs770504584 Chr.11: 69699300-69699300 on Build GRCh38
rs990188219 Chr.11: 69699327-69699327 on Build GRCh38
rs564723134 Chr.11: 69699169-69699169 on Build GRCh38
rs552560069 Chr.11: 69699144-69699144 on Build GRCh38
rs886599661 Chr.11: 69699208-69699208 on Build GRCh38
rs200919218 Chr.11: 69699247-69699247 on Build GRCh38
rs776547468 Chr.11: 69699301-69699301 on Build GRCh38
rs898180930 Chr.11: 69699199-69699199 on Build GRCh38
rs141765810 Chr.11: 69699312-69699312 on Build GRCh38
rs528016672 Chr.11: 69699134-69699134 on Build GRCh38
rs759246590 Chr.11: 69699307-69699307 on Build GRCh38
rs375312504 Chr.11: 69699334-69699334 on Build GRCh38
rs568253878 Chr.11: 69699207-69699207 on Build GRCh38
rs767740021 Chr.11: 69699331-69699331 on Build GRCh38

View Full Product Details