Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 22768328-22768589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF99
Entrez Gene ID
7652
Gene Name
zinc finger protein 99
Gene Aliases
C19orf9, F8281
UniGene
Hs.568380
Gene Chromosome Location
Chr.19: 22752183-22784171 on Build GRCh38
Transcripts
NM_001080409.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4076407 c.239G>A p.G80E Chr.19: 22768355-22768355 on Build GRCh38
474466 c.243A>G p.K81K Chr.19: 22768351-22768351 on Build GRCh38
6084783 c.196A>T p.I66F Chr.19: 22768398-22768398 on Build GRCh38
3531774 c.252G>A p.W84* Chr.19: 22768342-22768342 on Build GRCh38
6524824 c.200C>T p.A67V Chr.19: 22768394-22768394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147397251 Chr.19: 22768396-22768396 on Build GRCh38
rs767935662 Chr.19: 22768436-22768436 on Build GRCh38
rs888267311 Chr.19: 22768526-22768526 on Build GRCh38
rs192850061 Chr.19: 22768496-22768496 on Build GRCh38
rs777104611 Chr.19: 22768415-22768415 on Build GRCh38
rs566511080 Chr.19: 22768517-22768517 on Build GRCh38
rs542942085 Chr.19: 22768390-22768390 on Build GRCh38
rs771326518 Chr.19: 22768395-22768395 on Build GRCh38
rs761025659 Chr.19: 22768384-22768384 on Build GRCh38
rs562122209 Chr.19: 22768363-22768363 on Build GRCh38
rs957814127 Chr.19: 22768400-22768400 on Build GRCh38
rs185756090 Chr.19: 22768503-22768503 on Build GRCh38
rs749600953 Chr.19: 22768370-22768371 on Build GRCh38
rs759991432 Chr.19: 22768424-22768424 on Build GRCh38
rs773365626 Chr.19: 22768370-22768370 on Build GRCh38
rs777498366 Chr.19: 22768426-22768427 on Build GRCh38
rs1048258257 Chr.19: 22768521-22768521 on Build GRCh38
rs1046452841 Chr.19: 22768459-22768459 on Build GRCh38
rs755380643 Chr.19: 22768424-22768425 on Build GRCh38
rs949094408 Chr.19: 22768452-22768452 on Build GRCh38

View Full Product Details