Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150696782-150697051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM22
Entrez Gene ID
55696
Gene Name
RNA binding motif protein 22
Gene Aliases
Cwc2, ZC3H16, fSAP47
UniGene
Hs.713564
Gene Chromosome Location
Chr.5: 150690790-150701107 on Build GRCh38
Transcripts
NM_018047.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
449117 c.357G>A p.Q119Q Chr.5: 150696806-150696806 on Build GRCh38
4345757 c.142C>T p.R48C Chr.5: 150696874-150696874 on Build GRCh38
4813634 c.210G>A p.Q70Q Chr.5: 150696806-150696806 on Build GRCh38
4423229 c.308T>G p.F103C Chr.5: 150696855-150696855 on Build GRCh38
4423230 c.161T>G p.F54C Chr.5: 150696855-150696855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032446966 Chr.5: 150696906-150696906 on Build GRCh38
rs760431016 Chr.5: 150696902-150696902 on Build GRCh38
rs1012037824 Chr.5: 150696966-150696966 on Build GRCh38
rs769187224 Chr.5: 150696834-150696834 on Build GRCh38
rs780769552 Chr.5: 150696818-150696818 on Build GRCh38
rs747361840 Chr.5: 150696825-150696825 on Build GRCh38
rs753595942 Chr.5: 150696921-150696921 on Build GRCh38
rs545624171 Chr.5: 150696842-150696842 on Build GRCh38
rs758870393 Chr.5: 150696808-150696808 on Build GRCh38
rs776590022 Chr.5: 150696937-150696938 on Build GRCh38
rs764798661 Chr.5: 150696925-150696925 on Build GRCh38
rs779850042 Chr.5: 150696959-150696959 on Build GRCh38
rs970549830 Chr.5: 150696993-150696993 on Build GRCh38
rs773277046 Chr.5: 150696868-150696868 on Build GRCh38
rs746734791 Chr.5: 150696905-150696906 on Build GRCh38
rs749940276 Chr.5: 150696929-150696929 on Build GRCh38
rs974484887 Chr.5: 150696981-150696983 on Build GRCh38
rs761467818 Chr.5: 150696923-150696923 on Build GRCh38
rs531062098 Chr.5: 150696898-150696898 on Build GRCh38
rs763834304 Chr.5: 150696916-150696916 on Build GRCh38
rs150337870 Chr.5: 150696836-150696836 on Build GRCh38
rs528501769 Chr.5: 150696989-150696989 on Build GRCh38
rs766965555 Chr.5: 150696941-150696941 on Build GRCh38
rs886372153 Chr.5: 150696898-150696899 on Build GRCh38
rs905684357 Chr.5: 150696854-150696854 on Build GRCh38
rs192669869 Chr.5: 150697006-150697006 on Build GRCh38
rs41287132 Chr.5: 150696942-150696942 on Build GRCh38
rs41287134 Chr.5: 150696984-150696984 on Build GRCh38
rs765066694 Chr.5: 150696955-150696956 on Build GRCh38
rs377525329 Chr.5: 150697014-150697014 on Build GRCh38
rs34381493 Chr.5: 150696869-150696869 on Build GRCh38
rs955005313 Chr.5: 150696988-150696988 on Build GRCh38
rs771984728 Chr.5: 150696884-150696884 on Build GRCh38
rs1045154911 Chr.5: 150696850-150696850 on Build GRCh38
rs200975935 Chr.5: 150696844-150696844 on Build GRCh38
rs41290547 Chr.5: 150696994-150696994 on Build GRCh38
rs758036484 Chr.5: 150696940-150696940 on Build GRCh38
rs770870209 Chr.5: 150696932-150696932 on Build GRCh38
rs374202107 Chr.5: 150696918-150696918 on Build GRCh38

View Full Product Details