Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67307635-67307902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSH3
Entrez Gene ID
54961
Gene Name
slingshot protein phosphatase 3
Gene Aliases
SSH3L
UniGene
Hs.29173
Gene Chromosome Location
Chr.11: 67303448-67312607 on Build GRCh38
Transcripts
NM_017857.3 XM_017017942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7227777 c.714C>T p.S238S Chr.11: 67307660-67307660 on Build GRCh38
1492687 c.743C>T p.A248V Chr.11: 67307689-67307689 on Build GRCh38
7352955 c.829G>A p.A277T Chr.11: 67307883-67307883 on Build GRCh38
6070265 c.720G>A p.Q240Q Chr.11: 67307666-67307666 on Build GRCh38
6561622 c.797C>T p.S266L Chr.11: 67307851-67307851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138067329 Chr.11: 67307678-67307678 on Build GRCh38
rs745830198 Chr.11: 67307866-67307866 on Build GRCh38
rs866294085 Chr.11: 67307806-67307806 on Build GRCh38
rs57238619 Chr.11: 67307660-67307660 on Build GRCh38
rs376692542 Chr.11: 67307670-67307670 on Build GRCh38
rs758120069 Chr.11: 67307842-67307842 on Build GRCh38
rs767472087 Chr.11: 67307673-67307673 on Build GRCh38
rs148649808 Chr.11: 67307733-67307733 on Build GRCh38
rs779880796 Chr.11: 67307683-67307683 on Build GRCh38
rs61731782 Chr.11: 67307736-67307736 on Build GRCh38
rs778661828 Chr.11: 67307738-67307738 on Build GRCh38
rs759418319 Chr.11: 67307833-67307833 on Build GRCh38
rs774415123 Chr.11: 67307810-67307810 on Build GRCh38
rs370832879 Chr.11: 67307880-67307880 on Build GRCh38
rs763148055 Chr.11: 67307796-67307796 on Build GRCh38
rs376566798 Chr.11: 67307814-67307814 on Build GRCh38
rs376154199 Chr.11: 67307746-67307746 on Build GRCh38
rs776767490 Chr.11: 67307657-67307657 on Build GRCh38
rs1001398891 Chr.11: 67307714-67307714 on Build GRCh38
rs777408862 Chr.11: 67307765-67307765 on Build GRCh38
rs747586968 Chr.11: 67307766-67307766 on Build GRCh38
rs778369158 Chr.11: 67307687-67307687 on Build GRCh38
rs79688961 Chr.11: 67307726-67307726 on Build GRCh38
rs771481302 Chr.11: 67307704-67307704 on Build GRCh38
rs997669346 Chr.11: 67307716-67307716 on Build GRCh38
rs548768309 Chr.11: 67307710-67307710 on Build GRCh38
rs775829906 Chr.11: 67307789-67307789 on Build GRCh38
rs957572203 Chr.11: 67307849-67307849 on Build GRCh38
rs887621192 Chr.11: 67307727-67307727 on Build GRCh38
rs756680076 Chr.11: 67307875-67307875 on Build GRCh38
rs768787141 Chr.11: 67307799-67307799 on Build GRCh38
rs752460257 Chr.11: 67307839-67307839 on Build GRCh38
rs777051119 Chr.11: 67307706-67307706 on Build GRCh38
rs766359265 Chr.11: 67307818-67307818 on Build GRCh38
rs750413460 Chr.11: 67307731-67307731 on Build GRCh38
rs771671434 Chr.11: 67307696-67307696 on Build GRCh38
rs752829057 Chr.11: 67307661-67307661 on Build GRCh38
rs775533885 Chr.11: 67307713-67307713 on Build GRCh38
rs776660117 Chr.11: 67307836-67307836 on Build GRCh38
rs372060558 Chr.11: 67307788-67307788 on Build GRCh38
rs148268386 Chr.11: 67307686-67307686 on Build GRCh38
rs780388866 Chr.11: 67307881-67307881 on Build GRCh38
rs7114712 Chr.11: 67307662-67307662 on Build GRCh38
rs755951583 Chr.11: 67307681-67307681 on Build GRCh38
rs771914107 Chr.11: 67307697-67307697 on Build GRCh38
rs373944304 Chr.11: 67307721-67307721 on Build GRCh38
rs149465975 Chr.11: 67307712-67307712 on Build GRCh38
rs759791635 Chr.11: 67307659-67307659 on Build GRCh38
rs367831438 Chr.11: 67307723-67307723 on Build GRCh38
rs753680737 Chr.11: 67307826-67307826 on Build GRCh38
rs879134443 Chr.11: 67307792-67307792 on Build GRCh38
rs753677593 Chr.11: 67307735-67307735 on Build GRCh38
rs770022321 Chr.11: 67307786-67307786 on Build GRCh38
rs747717938 Chr.11: 67307691-67307691 on Build GRCh38
rs771474759 Chr.11: 67307767-67307767 on Build GRCh38
rs748956056 Chr.11: 67307685-67307685 on Build GRCh38
rs201934241 Chr.11: 67307876-67307876 on Build GRCh38
rs752370350 Chr.11: 67307743-67307743 on Build GRCh38
rs746337610 Chr.11: 67307878-67307878 on Build GRCh38
rs751129687 Chr.11: 67307867-67307867 on Build GRCh38

View Full Product Details