Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2250262-2250497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMH
Entrez Gene ID
268
Gene Name
anti-Mullerian hormone
Gene Aliases
MIF, MIS
UniGene
Hs.112432
Gene Chromosome Location
Chr.19: 2249114-2252073 on Build GRCh38
Transcripts
NM_000479.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5691309 c.451C>T p.P151S Chr.19: 2250375-2250375 on Build GRCh38
5660849 c.475C>T p.P159S Chr.19: 2250399-2250399 on Build GRCh38
5706940 c.484G>A p.A162T Chr.19: 2250408-2250408 on Build GRCh38
1392112 c.509C>A p.P170H Chr.19: 2250433-2250433 on Build GRCh38
3756561 c.546G>A p.P182P Chr.19: 2250470-2250470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751618322 Chr.19: 2250328-2250328 on Build GRCh38
rs769467742 Chr.19: 2250349-2250349 on Build GRCh38
rs771599497 Chr.19: 2250290-2250290 on Build GRCh38
rs773178911 Chr.19: 2250385-2250385 on Build GRCh38
rs17854573 Chr.19: 2250470-2250470 on Build GRCh38
rs540520333 Chr.19: 2250327-2250327 on Build GRCh38
rs201367909 Chr.19: 2250362-2250362 on Build GRCh38
rs760631522 Chr.19: 2250294-2250294 on Build GRCh38
rs773995807 Chr.19: 2250286-2250286 on Build GRCh38
rs768510419 Chr.19: 2250295-2250295 on Build GRCh38
rs936428725 Chr.19: 2250397-2250397 on Build GRCh38
rs565075895 Chr.19: 2250347-2250347 on Build GRCh38
rs374418184 Chr.19: 2250303-2250303 on Build GRCh38
rs774912206 Chr.19: 2250292-2250292 on Build GRCh38
rs753191322 Chr.19: 2250399-2250399 on Build GRCh38
rs866519862 Chr.19: 2250414-2250414 on Build GRCh38
rs768585333 Chr.19: 2250370-2250370 on Build GRCh38
rs749049690 Chr.19: 2250472-2250472 on Build GRCh38
rs1020151496 Chr.19: 2250299-2250299 on Build GRCh38
rs748063248 Chr.19: 2250454-2250454 on Build GRCh38
rs951526274 Chr.19: 2250354-2250354 on Build GRCh38
rs750657208 Chr.19: 2250316-2250316 on Build GRCh38
rs754233174 Chr.19: 2250409-2250409 on Build GRCh38
rs894766012 Chr.19: 2250284-2250284 on Build GRCh38
rs762070367 Chr.19: 2250449-2250450 on Build GRCh38
rs907222158 Chr.19: 2250432-2250432 on Build GRCh38
rs371874189 Chr.19: 2250424-2250424 on Build GRCh38
rs187807834 Chr.19: 2250396-2250396 on Build GRCh38
rs781564146 Chr.19: 2250333-2250333 on Build GRCh38
rs374588581 Chr.19: 2250390-2250390 on Build GRCh38
rs763192254 Chr.19: 2250388-2250388 on Build GRCh38
rs756662915 Chr.19: 2250402-2250402 on Build GRCh38
rs941604570 Chr.19: 2250435-2250435 on Build GRCh38
rs867455699 Chr.19: 2250400-2250400 on Build GRCh38
rs780894513 Chr.19: 2250436-2250436 on Build GRCh38
rs543640330 Chr.19: 2250358-2250358 on Build GRCh38
rs1024315749 Chr.19: 2250286-2250293 on Build GRCh38
rs774592796 Chr.19: 2250388-2250401 on Build GRCh38
rs975692513 Chr.19: 2250444-2250444 on Build GRCh38
rs958738596 Chr.19: 2250340-2250340 on Build GRCh38
rs776523141 Chr.19: 2250301-2250301 on Build GRCh38
rs752991250 Chr.19: 2250338-2250338 on Build GRCh38
rs751597455 Chr.19: 2250394-2250394 on Build GRCh38
rs554790862 Chr.19: 2250285-2250285 on Build GRCh38
rs763313469 Chr.19: 2250322-2250322 on Build GRCh38
rs771662423 Chr.19: 2250356-2250356 on Build GRCh38
rs183367973 Chr.19: 2250313-2250313 on Build GRCh38
rs778082272 Chr.19: 2250404-2250404 on Build GRCh38
rs529706314 Chr.19: 2250461-2250461 on Build GRCh38
rs367783341 Chr.19: 2250395-2250395 on Build GRCh38
rs745589165 Chr.19: 2250289-2250289 on Build GRCh38
rs768050453 Chr.19: 2250426-2250426 on Build GRCh38
rs370532523 Chr.19: 2250375-2250375 on Build GRCh38
rs779588404 Chr.19: 2250425-2250425 on Build GRCh38
rs139265145 Chr.19: 2250352-2250352 on Build GRCh38
rs1045987276 Chr.19: 2250432-2250432 on Build GRCh38
rs371965734 Chr.19: 2250379-2250379 on Build GRCh38
rs767836187 Chr.19: 2250475-2250475 on Build GRCh38
rs200523942 Chr.19: 2250477-2250477 on Build GRCh38

View Full Product Details