Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 55470125-55470390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT5A
Entrez Gene ID
7474
Gene Name
Wnt family member 5A
Gene Aliases
hWNT5A
UniGene
Hs.643085
Gene Chromosome Location
Chr.3: 55465715-55505261 on Build GRCh38
Transcripts
NM_001256105.1 NM_003392.4 XM_006713324.1 XM_011534085.2 XM_011534086.2 XM_011534087.2 XM_011534088.2 XM_011534089.1 XM_017007127.1 XM_017007128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
192113 c.1207G>A p.V403M Chr.3: 55470307-55470307 on Build GRCh38
6823264 c.1335G>A p.T445T Chr.3: 55470179-55470179 on Build GRCh38
420340 c.1372G>C p.V458L Chr.3: 55470142-55470142 on Build GRCh38
6823263 c.1304G>A p.R435H Chr.3: 55470210-55470210 on Build GRCh38
2781236 c.1244G>A p.G415D Chr.3: 55470270-55470270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754080980 Chr.3: 55470179-55470179 on Build GRCh38
rs770199302 Chr.3: 55470331-55470331 on Build GRCh38
rs755247138 Chr.3: 55470285-55470285 on Build GRCh38
rs779036055 Chr.3: 55470187-55470187 on Build GRCh38
rs531456992 Chr.3: 55470332-55470332 on Build GRCh38
rs768254997 Chr.3: 55470241-55470241 on Build GRCh38
rs946840827 Chr.3: 55470273-55470273 on Build GRCh38
rs573691468 Chr.3: 55470199-55470199 on Build GRCh38
rs762576376 Chr.3: 55470230-55470230 on Build GRCh38
rs755298573 Chr.3: 55470180-55470180 on Build GRCh38
rs747860258 Chr.3: 55470337-55470337 on Build GRCh38
rs747485846 Chr.3: 55470310-55470310 on Build GRCh38
rs773819361 Chr.3: 55470333-55470333 on Build GRCh38
rs772896188 Chr.3: 55470346-55470346 on Build GRCh38
rs747534689 Chr.3: 55470211-55470211 on Build GRCh38
rs746302333 Chr.3: 55470329-55470329 on Build GRCh38
rs757813200 Chr.3: 55470324-55470324 on Build GRCh38
rs375700896 Chr.3: 55470249-55470249 on Build GRCh38
rs767068436 Chr.3: 55470253-55470253 on Build GRCh38
rs369701725 Chr.3: 55470298-55470298 on Build GRCh38
rs544912525 Chr.3: 55470353-55470353 on Build GRCh38
rs752974227 Chr.3: 55470303-55470303 on Build GRCh38
rs750281702 Chr.3: 55470255-55470255 on Build GRCh38
rs777938109 Chr.3: 55470307-55470307 on Build GRCh38
rs961757574 Chr.3: 55470251-55470251 on Build GRCh38
rs542775163 Chr.3: 55470229-55470229 on Build GRCh38
rs755916316 Chr.3: 55470155-55470155 on Build GRCh38
rs771445451 Chr.3: 55470343-55470343 on Build GRCh38
rs201975128 Chr.3: 55470252-55470252 on Build GRCh38
rs781527130 Chr.3: 55470328-55470328 on Build GRCh38
rs759194223 Chr.3: 55470357-55470357 on Build GRCh38
rs1050351761 Chr.3: 55470311-55470311 on Build GRCh38
rs771248572 Chr.3: 55470228-55470228 on Build GRCh38
rs372423949 Chr.3: 55470188-55470188 on Build GRCh38
rs760675240 Chr.3: 55470348-55470348 on Build GRCh38
rs931674113 Chr.3: 55470291-55470291 on Build GRCh38
rs1044254835 Chr.3: 55470320-55470320 on Build GRCh38
rs373282952 Chr.3: 55470259-55470259 on Build GRCh38
rs900039147 Chr.3: 55470349-55470349 on Build GRCh38
rs766254001 Chr.3: 55470284-55470284 on Build GRCh38
rs776714857 Chr.3: 55470356-55470356 on Build GRCh38
rs765101926 Chr.3: 55470366-55470366 on Build GRCh38
rs922958622 Chr.3: 55470276-55470276 on Build GRCh38
rs780019128 Chr.3: 55470162-55470162 on Build GRCh38
rs758357805 Chr.3: 55470306-55470306 on Build GRCh38

View Full Product Details