Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 82365020-82365251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPBG
Entrez Gene ID
7162
Gene Name
trophoblast glycoprotein
Gene Aliases
5T4, 5T4AG, M6P1, WAIF1
UniGene
Hs.71947
Gene Chromosome Location
Chr.6: 82363206-82370828 on Build GRCh38
Transcripts
NM_001166392.1 NM_006670.4 XM_011536097.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4984237 c.131C>T p.S44L Chr.6: 82365092-82365092 on Build GRCh38
6207799 c.105C>A p.T35T Chr.6: 82365066-82365066 on Build GRCh38
5794086 c.217C>G p.R73G Chr.6: 82365178-82365178 on Build GRCh38
5061966 c.218G>A p.R73H Chr.6: 82365179-82365179 on Build GRCh38
6346940 c.138G>T p.P46P Chr.6: 82365099-82365099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201739068 Chr.6: 82365211-82365211 on Build GRCh38
rs561305359 Chr.6: 82365046-82365046 on Build GRCh38
rs755457753 Chr.6: 82365233-82365233 on Build GRCh38
rs764801081 Chr.6: 82365139-82365139 on Build GRCh38
rs898949683 Chr.6: 82365096-82365096 on Build GRCh38
rs771809173 Chr.6: 82365112-82365112 on Build GRCh38
rs780454043 Chr.6: 82365087-82365087 on Build GRCh38
rs774506603 Chr.6: 82365203-82365203 on Build GRCh38
rs139823445 Chr.6: 82365111-82365111 on Build GRCh38
rs753716772 Chr.6: 82365134-82365134 on Build GRCh38
rs532320183 Chr.6: 82365210-82365210 on Build GRCh38
rs1051351192 Chr.6: 82365066-82365066 on Build GRCh38
rs769819568 Chr.6: 82365179-82365179 on Build GRCh38
rs753377946 Chr.6: 82365068-82365068 on Build GRCh38
rs1014200166 Chr.6: 82365144-82365144 on Build GRCh38
rs752008376 Chr.6: 82365148-82365148 on Build GRCh38
rs150807599 Chr.6: 82365219-82365219 on Build GRCh38
rs984020597 Chr.6: 82365089-82365089 on Build GRCh38
rs530126449 Chr.6: 82365060-82365060 on Build GRCh38
rs922779150 Chr.6: 82365231-82365231 on Build GRCh38
rs778127024 Chr.6: 82365081-82365083 on Build GRCh38
rs776616045 Chr.6: 82365208-82365208 on Build GRCh38
rs780608227 Chr.6: 82365145-82365145 on Build GRCh38
rs781578437 Chr.6: 82365186-82365188 on Build GRCh38
rs778444688 Chr.6: 82365072-82365072 on Build GRCh38
rs755349428 Chr.6: 82365150-82365150 on Build GRCh38
rs764040683 Chr.6: 82365128-82365128 on Build GRCh38
rs757338696 Chr.6: 82365177-82365183 on Build GRCh38
rs756022835 Chr.6: 82365042-82365042 on Build GRCh38
rs758758325 Chr.6: 82365080-82365080 on Build GRCh38
rs748263652 Chr.6: 82365102-82365102 on Build GRCh38
rs777884045 Chr.6: 82365187-82365187 on Build GRCh38
rs769841559 Chr.6: 82365105-82365105 on Build GRCh38
rs967351830 Chr.6: 82365198-82365198 on Build GRCh38
rs922844319 Chr.6: 82365058-82365058 on Build GRCh38
rs976020447 Chr.6: 82365055-82365055 on Build GRCh38
rs144253219 Chr.6: 82365220-82365220 on Build GRCh38
rs143139849 Chr.6: 82365199-82365199 on Build GRCh38
rs749510960 Chr.6: 82365194-82365194 on Build GRCh38
rs371857715 Chr.6: 82365082-82365082 on Build GRCh38
rs773369774 Chr.6: 82365109-82365109 on Build GRCh38
rs761235105 Chr.6: 82365138-82365138 on Build GRCh38
rs758759402 Chr.6: 82365052-82365054 on Build GRCh38
rs760475820 Chr.6: 82365205-82365205 on Build GRCh38
rs1054837351 Chr.6: 82365155-82365155 on Build GRCh38
rs751203253 Chr.6: 82365228-82365228 on Build GRCh38
rs775233830 Chr.6: 82365115-82365115 on Build GRCh38
rs565218547 Chr.6: 82365099-82365099 on Build GRCh38
rs769256988 Chr.6: 82365097-82365097 on Build GRCh38
rs1037437585 Chr.6: 82365083-82365083 on Build GRCh38
rs758049755 Chr.6: 82365141-82365141 on Build GRCh38
rs1045931571 Chr.6: 82365132-82365132 on Build GRCh38
rs895698606 Chr.6: 82365119-82365119 on Build GRCh38
rs747096492 Chr.6: 82365084-82365086 on Build GRCh38
rs760420645 Chr.6: 82365120-82365120 on Build GRCh38
rs747570726 Chr.6: 82365092-82365092 on Build GRCh38
rs371334832 Chr.6: 82365073-82365073 on Build GRCh38
rs934246054 Chr.6: 82365061-82365061 on Build GRCh38

View Full Product Details