Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 24208149-24208412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFX
Entrez Gene ID
7543
Gene Name
zinc finger protein, X-linked
Gene Aliases
ZNF926
UniGene
Hs.336681
Gene Chromosome Location
Chr.X: 24148994-24216255 on Build GRCh38
Transcripts
NM_001178084.1 NM_001178085.1 NM_001178086.1 NM_001178095.1 NM_003410.3 XM_005274591.4 XM_005274592.3 XM_005274594.4 XM_006724513.3 XM_006724515.3 XM_011545578.1 XM_011545579.1 XM_011545581.2 XM_011545583.2 XM_017029788.1 XM_017029789.1 XM_017029790.1 XM_017029791.1 XM_017029792.1 XM_017029793.1 XM_017029794.1 XM_017029795.1 XM_017029796.1 XM_017029797.1 XM_017029798.1 XM_017029799.1 XM_017029800.1 XM_017029801.1

SNP Detail

SNP ID Chromosome Location
rs1062608 Chr.X: 24208289-24208289 on Build GRCh38
rs747079975 Chr.X: 24208312-24208312 on Build GRCh38
rs780034453 Chr.X: 24208301-24208301 on Build GRCh38
rs768177592 Chr.X: 24208279-24208279 on Build GRCh38
rs777201713 Chr.X: 24208202-24208202 on Build GRCh38
rs767333577 Chr.X: 24208210-24208212 on Build GRCh38
rs762255489 Chr.X: 24208207-24208207 on Build GRCh38
rs762272041 Chr.X: 24208338-24208338 on Build GRCh38
rs375874182 Chr.X: 24208352-24208352 on Build GRCh38
rs146342075 Chr.X: 24208294-24208294 on Build GRCh38
rs761497679 Chr.X: 24208297-24208297 on Build GRCh38
rs769342733 Chr.X: 24208313-24208313 on Build GRCh38
rs766051923 Chr.X: 24208386-24208386 on Build GRCh38
rs765820465 Chr.X: 24208273-24208274 on Build GRCh38
rs765559608 Chr.X: 24208327-24208327 on Build GRCh38
rs375157373 Chr.X: 24208287-24208287 on Build GRCh38
rs776080622 Chr.X: 24208281-24208281 on Build GRCh38
rs199548791 Chr.X: 24208295-24208295 on Build GRCh38
rs765857452 Chr.X: 24208225-24208225 on Build GRCh38
rs943473548 Chr.X: 24208240-24208240 on Build GRCh38
rs371795466 Chr.X: 24208348-24208348 on Build GRCh38
rs769418725 Chr.X: 24208339-24208341 on Build GRCh38
rs753340045 Chr.X: 24208277-24208282 on Build GRCh38
rs764956102 Chr.X: 24208212-24208212 on Build GRCh38
rs762548241 Chr.X: 24208214-24208214 on Build GRCh38
rs997542526 Chr.X: 24208232-24208232 on Build GRCh38
rs762555522 Chr.X: 24208379-24208379 on Build GRCh38
rs760304619 Chr.X: 24208273-24208275 on Build GRCh38
rs746865044 Chr.X: 24208213-24208213 on Build GRCh38
rs754978527 Chr.X: 24208255-24208255 on Build GRCh38
rs755842356 Chr.X: 24208293-24208293 on Build GRCh38
rs751040155 Chr.X: 24208237-24208237 on Build GRCh38
rs142016517 Chr.X: 24208288-24208288 on Build GRCh38
rs772312837 Chr.X: 24208298-24208298 on Build GRCh38
rs750036944 Chr.X: 24208272-24208273 on Build GRCh38

View Full Product Details