Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95737010-95737261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOBTB3
Entrez Gene ID
22836
Gene Name
Rho related BTB domain containing 3
Gene Aliases
-
UniGene
Hs.445030
Gene Chromosome Location
Chr.5: 95713522-95796367 on Build GRCh38
Transcripts
NM_014899.3 XM_011543279.2 XM_017009236.1 XM_017009237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6766247 c.415+1G>T p.? Chr.5: 95737076-95737076 on Build GRCh38
6383404 c.392C>A p.A131D Chr.5: 95737052-95737052 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs947814871 Chr.5: 95737142-95737142 on Build GRCh38
rs777599710 Chr.5: 95737070-95737070 on Build GRCh38
rs966476082 Chr.5: 95737208-95737208 on Build GRCh38
rs1014935239 Chr.5: 95737201-95737201 on Build GRCh38
rs746892692 Chr.5: 95737106-95737106 on Build GRCh38
rs188272478 Chr.5: 95737229-95737229 on Build GRCh38
rs1004852081 Chr.5: 95737191-95737191 on Build GRCh38
rs898350461 Chr.5: 95737200-95737200 on Build GRCh38
rs925674516 Chr.5: 95737230-95737230 on Build GRCh38
rs776872396 Chr.5: 95737092-95737093 on Build GRCh38
rs1021742111 Chr.5: 95737231-95737231 on Build GRCh38
rs952121627 Chr.5: 95737169-95737169 on Build GRCh38
rs953549534 Chr.5: 95737221-95737221 on Build GRCh38
rs979585696 Chr.5: 95737228-95737228 on Build GRCh38
rs1044845818 Chr.5: 95737170-95737170 on Build GRCh38
rs116353007 Chr.5: 95737211-95737211 on Build GRCh38
rs747999813 Chr.5: 95737068-95737068 on Build GRCh38
rs778707106 Chr.5: 95737052-95737052 on Build GRCh38
rs1056172205 Chr.5: 95737124-95737124 on Build GRCh38
rs1001795379 Chr.5: 95737129-95737129 on Build GRCh38
rs771941215 Chr.5: 95737069-95737069 on Build GRCh38
rs143245429 Chr.5: 95737216-95737216 on Build GRCh38
rs1027717152 Chr.5: 95737137-95737137 on Build GRCh38
rs571722767 Chr.5: 95737121-95737121 on Build GRCh38
rs976350082 Chr.5: 95737210-95737210 on Build GRCh38

View Full Product Details