Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48137161-48137420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKAP1
Entrez Gene ID
8631
Gene Name
src kinase associated phosphoprotein 1
Gene Aliases
HEL-S-81p, SCAP1, SKAP55
UniGene
Hs.316931
Gene Chromosome Location
Chr.17: 48133438-48430291 on Build GRCh38
Transcripts
NM_001075099.1 NM_003726.3 XM_005257755.3 XM_017025257.1 XM_017025258.1 XM_017025259.1 XM_017025260.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7389999 c.980A>G p.E327G Chr.17: 48137336-48137336 on Build GRCh38
4268270 c.1048C>T p.L350F Chr.17: 48137268-48137268 on Build GRCh38
7390000 c.979G>A p.E327K Chr.17: 48137337-48137337 on Build GRCh38
6839450 c.1044G>T p.E348D Chr.17: 48137272-48137272 on Build GRCh38
5621822 c.986A>G p.N329S Chr.17: 48137330-48137330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932820063 Chr.17: 48137255-48137255 on Build GRCh38
rs143407064 Chr.17: 48137291-48137291 on Build GRCh38
rs761632125 Chr.17: 48137279-48137279 on Build GRCh38
rs867194496 Chr.17: 48137237-48137237 on Build GRCh38
rs931190101 Chr.17: 48137388-48137388 on Build GRCh38
rs772095586 Chr.17: 48137250-48137250 on Build GRCh38
rs911791440 Chr.17: 48137195-48137195 on Build GRCh38
rs767201665 Chr.17: 48137189-48137189 on Build GRCh38
rs749068512 Chr.17: 48137191-48137191 on Build GRCh38
rs778264300 Chr.17: 48137260-48137260 on Build GRCh38
rs767395301 Chr.17: 48137327-48137327 on Build GRCh38
rs748826699 Chr.17: 48137234-48137234 on Build GRCh38
rs781698343 Chr.17: 48137381-48137381 on Build GRCh38
rs773969850 Chr.17: 48137363-48137364 on Build GRCh38
rs774415489 Chr.17: 48137312-48137312 on Build GRCh38
rs79874741 Chr.17: 48137187-48137187 on Build GRCh38
rs375826607 Chr.17: 48137377-48137377 on Build GRCh38
rs750132050 Chr.17: 48137213-48137213 on Build GRCh38
rs373548020 Chr.17: 48137270-48137270 on Build GRCh38
rs535417046 Chr.17: 48137196-48137196 on Build GRCh38
rs761625794 Chr.17: 48137316-48137316 on Build GRCh38
rs369814528 Chr.17: 48137192-48137192 on Build GRCh38
rs770175322 Chr.17: 48137292-48137292 on Build GRCh38
rs1037096063 Chr.17: 48137362-48137362 on Build GRCh38
rs770754098 Chr.17: 48137351-48137352 on Build GRCh38
rs766023758 Chr.17: 48137361-48137361 on Build GRCh38
rs775791999 Chr.17: 48137293-48137293 on Build GRCh38
rs746227109 Chr.17: 48137389-48137389 on Build GRCh38
rs755897488 Chr.17: 48137360-48137360 on Build GRCh38
rs764258407 Chr.17: 48137308-48137308 on Build GRCh38
rs779633634 Chr.17: 48137219-48137219 on Build GRCh38
rs750208164 Chr.17: 48137347-48137347 on Build GRCh38
rs540968106 Chr.17: 48137368-48137368 on Build GRCh38
rs74885375 Chr.17: 48137374-48137374 on Build GRCh38
rs747589246 Chr.17: 48137375-48137375 on Build GRCh38
rs763142121 Chr.17: 48137296-48137296 on Build GRCh38
rs755700577 Chr.17: 48137215-48137215 on Build GRCh38
rs754416960 Chr.17: 48137251-48137251 on Build GRCh38

View Full Product Details