Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52016088-52016337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF614
Entrez Gene ID
80110
Gene Name
zinc finger protein 614
Gene Aliases
-
UniGene
Hs.292336
Gene Chromosome Location
Chr.19: 52013324-52028427 on Build GRCh38
Transcripts
NM_025040.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4080774 c.1456A>G p.T486A Chr.19: 52016142-52016142 on Build GRCh38
1000316 c.1445C>T p.P482L Chr.19: 52016153-52016153 on Build GRCh38
1612687 c.1424G>C p.R475T Chr.19: 52016174-52016174 on Build GRCh38
7166278 c.1398G>C p.Q466H Chr.19: 52016200-52016200 on Build GRCh38
6085534 c.1312A>G p.T438A Chr.19: 52016286-52016286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772956705 Chr.19: 52016238-52016238 on Build GRCh38
rs772321022 Chr.19: 52016123-52016123 on Build GRCh38
rs927021321 Chr.19: 52016117-52016117 on Build GRCh38
rs1050506097 Chr.19: 52016194-52016194 on Build GRCh38
rs762724681 Chr.19: 52016146-52016146 on Build GRCh38
rs760696985 Chr.19: 52016182-52016196 on Build GRCh38
rs752186759 Chr.19: 52016179-52016179 on Build GRCh38
rs759212450 Chr.19: 52016167-52016167 on Build GRCh38
rs143573470 Chr.19: 52016182-52016182 on Build GRCh38
rs148053965 Chr.19: 52016208-52016208 on Build GRCh38
rs762182085 Chr.19: 52016142-52016145 on Build GRCh38
rs766977365 Chr.19: 52016170-52016170 on Build GRCh38
rs373361966 Chr.19: 52016203-52016203 on Build GRCh38
rs766575145 Chr.19: 52016270-52016270 on Build GRCh38
rs773459814 Chr.19: 52016134-52016134 on Build GRCh38
rs749265951 Chr.19: 52016259-52016259 on Build GRCh38
rs369045441 Chr.19: 52016139-52016139 on Build GRCh38
rs754396424 Chr.19: 52016296-52016296 on Build GRCh38
rs773413455 Chr.19: 52016148-52016149 on Build GRCh38
rs767643393 Chr.19: 52016145-52016146 on Build GRCh38
rs750791115 Chr.19: 52016189-52016189 on Build GRCh38
rs755280234 Chr.19: 52016225-52016225 on Build GRCh38
rs747126979 Chr.19: 52016285-52016285 on Build GRCh38
rs779342561 Chr.19: 52016185-52016185 on Build GRCh38
rs201885469 Chr.19: 52016119-52016119 on Build GRCh38
rs768796873 Chr.19: 52016133-52016133 on Build GRCh38
rs780489737 Chr.19: 52016124-52016124 on Build GRCh38
rs200985913 Chr.19: 52016140-52016140 on Build GRCh38
rs770668249 Chr.19: 52016265-52016265 on Build GRCh38
rs879049514 Chr.19: 52016172-52016172 on Build GRCh38
rs369503299 Chr.19: 52016287-52016287 on Build GRCh38
rs766428658 Chr.19: 52016267-52016269 on Build GRCh38
rs73934727 Chr.19: 52016221-52016221 on Build GRCh38
rs115605807 Chr.19: 52016164-52016164 on Build GRCh38
rs376539129 Chr.19: 52016234-52016234 on Build GRCh38
rs568147214 Chr.19: 52016273-52016273 on Build GRCh38
rs766169981 Chr.19: 52016151-52016151 on Build GRCh38
rs758623266 Chr.19: 52016193-52016193 on Build GRCh38
rs375760839 Chr.19: 52016120-52016120 on Build GRCh38
rs767204005 Chr.19: 52016300-52016300 on Build GRCh38
rs746264178 Chr.19: 52016122-52016122 on Build GRCh38
rs769766064 Chr.19: 52016236-52016236 on Build GRCh38
rs747239289 Chr.19: 52016125-52016125 on Build GRCh38
rs549653554 Chr.19: 52016235-52016235 on Build GRCh38
rs751657563 Chr.19: 52016295-52016295 on Build GRCh38
rs757738536 Chr.19: 52016113-52016113 on Build GRCh38
rs946330118 Chr.19: 52016144-52016144 on Build GRCh38
rs201085926 Chr.19: 52016239-52016239 on Build GRCh38
rs771004501 Chr.19: 52016294-52016294 on Build GRCh38

View Full Product Details