Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 77583350-77583605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN12
Entrez Gene ID
5782
Gene Name
protein tyrosine phosphatase, non-receptor type 12
Gene Aliases
PTP-PEST, PTPG1
UniGene
Hs.61812
Gene Chromosome Location
Chr.7: 77537160-77640071 on Build GRCh38
Transcripts
NM_001131008.1 NM_001131009.1 NM_002835.3 XM_005250518.2 XM_006716073.3 XM_011516444.1 XM_017012474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3640727 c.300A>C p.P100P Chr.7: 77583569-77583569 on Build GRCh38
3882504 c.319C>G p.Q107E Chr.7: 77583588-77583588 on Build GRCh38
7329220 c.317C>T p.T106I Chr.7: 77583586-77583586 on Build GRCh38
4541695 c.304G>T p.A102S Chr.7: 77583573-77583573 on Build GRCh38
297072 c.292T>C p.Y98H Chr.7: 77583561-77583561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192203807 Chr.7: 77583578-77583578 on Build GRCh38
rs559269943 Chr.7: 77583573-77583573 on Build GRCh38
rs554163203 Chr.7: 77583461-77583461 on Build GRCh38
rs769543464 Chr.7: 77583494-77583494 on Build GRCh38
rs956831122 Chr.7: 77583383-77583383 on Build GRCh38
rs571288211 Chr.7: 77583415-77583415 on Build GRCh38
rs149279587 Chr.7: 77583562-77583562 on Build GRCh38
rs987364425 Chr.7: 77583487-77583487 on Build GRCh38
rs1048477188 Chr.7: 77583424-77583424 on Build GRCh38
rs1032100739 Chr.7: 77583421-77583421 on Build GRCh38
rs567899909 Chr.7: 77583501-77583501 on Build GRCh38
rs182097847 Chr.7: 77583390-77583390 on Build GRCh38
rs536552312 Chr.7: 77583566-77583566 on Build GRCh38
rs369228581 Chr.7: 77583557-77583557 on Build GRCh38
rs968997827 Chr.7: 77583527-77583527 on Build GRCh38
rs768338922 Chr.7: 77583524-77583524 on Build GRCh38
rs1040134244 Chr.7: 77583403-77583403 on Build GRCh38
rs912842082 Chr.7: 77583387-77583387 on Build GRCh38
rs188868568 Chr.7: 77583429-77583429 on Build GRCh38
rs904877698 Chr.7: 77583414-77583414 on Build GRCh38
rs958061973 Chr.7: 77583428-77583428 on Build GRCh38
rs776245246 Chr.7: 77583551-77583551 on Build GRCh38
rs758214649 Chr.7: 77583511-77583511 on Build GRCh38
rs1023739508 Chr.7: 77583506-77583506 on Build GRCh38
rs536395499 Chr.7: 77583434-77583434 on Build GRCh38
rs770147304 Chr.7: 77583558-77583558 on Build GRCh38
rs988324391 Chr.7: 77583384-77583384 on Build GRCh38
rs375752041 Chr.7: 77583522-77583522 on Build GRCh38
rs746464188 Chr.7: 77583523-77583523 on Build GRCh38
rs944290965 Chr.7: 77583393-77583393 on Build GRCh38
rs975850491 Chr.7: 77583537-77583537 on Build GRCh38
rs931401652 Chr.7: 77583422-77583422 on Build GRCh38

View Full Product Details