Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 878160-878427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT4
Entrez Gene ID
51378
Gene Name
angiopoietin 4
Gene Aliases
ANG3, ANG4
UniGene
Hs.278973
Gene Chromosome Location
Chr.20: 869899-916367 on Build GRCh38
Transcripts
NM_001322809.1 NM_015985.3 XM_011529239.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1483864 c.1060G>A p.G354R Chr.20: 878321-878321 on Build GRCh38
6415419 c.1218C>A p.Y406* Chr.20: 878163-878163 on Build GRCh38
73655 c.1136G>A p.R379H Chr.20: 878245-878245 on Build GRCh38
6416737 c.1153T>G p.W385G Chr.20: 878228-878228 on Build GRCh38
2150444 c.1103A>T p.H368L Chr.20: 878278-878278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780368430 Chr.20: 878220-878220 on Build GRCh38
rs539498057 Chr.20: 878392-878392 on Build GRCh38
rs755197109 Chr.20: 878333-878333 on Build GRCh38
rs1010346349 Chr.20: 878258-878258 on Build GRCh38
rs778903378 Chr.20: 878255-878255 on Build GRCh38
rs745677751 Chr.20: 878272-878272 on Build GRCh38
rs181395936 Chr.20: 878344-878344 on Build GRCh38
rs35701717 Chr.20: 878378-878379 on Build GRCh38
rs910727621 Chr.20: 878403-878403 on Build GRCh38
rs373903775 Chr.20: 878204-878204 on Build GRCh38
rs937343140 Chr.20: 878235-878235 on Build GRCh38
rs549774453 Chr.20: 878351-878351 on Build GRCh38
rs748158262 Chr.20: 878246-878246 on Build GRCh38
rs773811714 Chr.20: 878321-878321 on Build GRCh38
rs200902475 Chr.20: 878372-878372 on Build GRCh38
rs200458141 Chr.20: 878320-878320 on Build GRCh38
rs976865019 Chr.20: 878314-878314 on Build GRCh38
rs756461495 Chr.20: 878217-878217 on Build GRCh38
rs958075097 Chr.20: 878216-878216 on Build GRCh38
rs370544958 Chr.20: 878327-878327 on Build GRCh38
rs768014934 Chr.20: 878211-878211 on Build GRCh38
rs573758146 Chr.20: 878283-878283 on Build GRCh38
rs370137040 Chr.20: 878307-878307 on Build GRCh38
rs148754514 Chr.20: 878247-878247 on Build GRCh38
rs112090528 Chr.20: 878322-878322 on Build GRCh38
rs745864356 Chr.20: 878355-878355 on Build GRCh38
rs753972032 Chr.20: 878259-878290 on Build GRCh38
rs755276380 Chr.20: 878336-878337 on Build GRCh38
rs929039300 Chr.20: 878385-878385 on Build GRCh38
rs765385171 Chr.20: 878343-878343 on Build GRCh38
rs769668731 Chr.20: 878280-878280 on Build GRCh38
rs201062169 Chr.20: 878199-878199 on Build GRCh38
rs775414137 Chr.20: 878282-878282 on Build GRCh38
rs749036353 Chr.20: 878371-878371 on Build GRCh38
rs908074894 Chr.20: 878183-878183 on Build GRCh38
rs762660025 Chr.20: 878196-878196 on Build GRCh38
rs869171 Chr.20: 878198-878198 on Build GRCh38
rs769728413 Chr.20: 878360-878383 on Build GRCh38
rs761338804 Chr.20: 878308-878308 on Build GRCh38
rs1018610446 Chr.20: 878331-878331 on Build GRCh38
rs201941980 Chr.20: 878188-878188 on Build GRCh38
rs370922444 Chr.20: 878359-878359 on Build GRCh38
rs750776952 Chr.20: 878212-878212 on Build GRCh38
rs768360241 Chr.20: 878299-878299 on Build GRCh38
rs758460998 Chr.20: 878348-878348 on Build GRCh38
rs890966005 Chr.20: 878315-878315 on Build GRCh38
rs748876652 Chr.20: 878276-878276 on Build GRCh38
rs766785366 Chr.20: 878326-878326 on Build GRCh38
rs201791557 Chr.20: 878245-878245 on Build GRCh38
rs754110274 Chr.20: 878238-878238 on Build GRCh38
rs780002725 Chr.20: 878368-878368 on Build GRCh38

View Full Product Details