Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 443593-443856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA11P
Entrez Gene ID
79963
Gene Name
ATP binding cassette subfamily A member 11, pseudogene
Gene Aliases
ABCA11, EST1133530
UniGene
Hs.677493
Gene Chromosome Location
Chr.4: 425435-474209 on Build GRCh38
Transcripts
NR_002451.2 NM_133474.3

Gene Symbol
ZNF721
Entrez Gene ID
170960
Gene Name
zinc finger protein 721
Gene Aliases
-
UniGene
Hs.428360
Gene Chromosome Location
Chr.4: 439984-499171 on Build GRCh38
Transcripts
NR_002451.2 NM_133474.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
587469 c.168T>C p.F56F Chr.4: 443609-443609 on Build GRCh38
6832689 c.27T>C p.H9H Chr.4: 443750-443750 on Build GRCh38
6330974 c.718G>A p.E240K Chr.4: 443749-443749 on Build GRCh38
7095955 c.771C>T p.V257V Chr.4: 443696-443696 on Build GRCh38
5978792 c.56A>G p.Y19C Chr.4: 443721-443721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782529181 Chr.4: 443752-443752 on Build GRCh38
rs372601849 Chr.4: 443822-443822 on Build GRCh38
rs782091479 Chr.4: 443743-443743 on Build GRCh38
rs782173767 Chr.4: 443706-443706 on Build GRCh38
rs374614076 Chr.4: 443647-443647 on Build GRCh38
rs782390093 Chr.4: 443787-443787 on Build GRCh38
rs782589467 Chr.4: 443631-443631 on Build GRCh38
rs782645432 Chr.4: 443764-443764 on Build GRCh38
rs782756351 Chr.4: 443747-443747 on Build GRCh38
rs781990772 Chr.4: 443684-443684 on Build GRCh38
rs781809624 Chr.4: 443817-443817 on Build GRCh38
rs200530483 Chr.4: 443754-443754 on Build GRCh38
rs371495220 Chr.4: 443648-443648 on Build GRCh38
rs1016552350 Chr.4: 443772-443772 on Build GRCh38
rs192018843 Chr.4: 443759-443759 on Build GRCh38
rs782245693 Chr.4: 443765-443765 on Build GRCh38
rs781896562 Chr.4: 443748-443748 on Build GRCh38
rs376631876 Chr.4: 443736-443736 on Build GRCh38
rs368280658 Chr.4: 443813-443813 on Build GRCh38
rs782507651 Chr.4: 443743-443743 on Build GRCh38
rs904349723 Chr.4: 443628-443628 on Build GRCh38
rs782729202 Chr.4: 443683-443683 on Build GRCh38
rs1023065306 Chr.4: 443751-443751 on Build GRCh38
rs781827787 Chr.4: 443733-443733 on Build GRCh38
rs781912933 Chr.4: 443669-443669 on Build GRCh38
rs782385298 Chr.4: 443716-443716 on Build GRCh38
rs782206149 Chr.4: 443769-443769 on Build GRCh38
rs374982400 Chr.4: 443692-443692 on Build GRCh38
rs781941325 Chr.4: 443712-443712 on Build GRCh38
rs376255771 Chr.4: 443621-443621 on Build GRCh38
rs782155360 Chr.4: 443814-443814 on Build GRCh38
rs782273290 Chr.4: 443653-443653 on Build GRCh38
rs199512799 Chr.4: 443632-443632 on Build GRCh38
rs782803961 Chr.4: 443710-443710 on Build GRCh38
rs782057251 Chr.4: 443714-443714 on Build GRCh38
rs1002436872 Chr.4: 443761-443761 on Build GRCh38
rs367900463 Chr.4: 443731-443731 on Build GRCh38
rs375487945 Chr.4: 443808-443808 on Build GRCh38
rs782669527 Chr.4: 443768-443768 on Build GRCh38
rs919308563 Chr.4: 443785-443785 on Build GRCh38
rs781979057 Chr.4: 443797-443797 on Build GRCh38
rs782315918 Chr.4: 443667-443667 on Build GRCh38
rs782084183 Chr.4: 443786-443786 on Build GRCh38
rs781972052 Chr.4: 443780-443780 on Build GRCh38
rs972680956 Chr.4: 443777-443777 on Build GRCh38
rs782418578 Chr.4: 443766-443766 on Build GRCh38
rs781989897 Chr.4: 443704-443704 on Build GRCh38
rs781793550 Chr.4: 443629-443629 on Build GRCh38
rs200907568 Chr.4: 443773-443773 on Build GRCh38
rs370185676 Chr.4: 443680-443680 on Build GRCh38

View Full Product Details