Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 214331991-214332224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMYD2
Entrez Gene ID
56950
Gene Name
SET and MYND domain containing 2
Gene Aliases
HSKM-B, KMT3C, ZMYND14
UniGene
Hs.66170
Gene Chromosome Location
Chr.1: 214281101-214337134 on Build GRCh38
Transcripts
NM_020197.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2151411 c.1032C>A p.Y344* Chr.1: 214332112-214332112 on Build GRCh38
5429812 c.1033C>G p.Q345E Chr.1: 214332113-214332113 on Build GRCh38
1295856 c.955G>A p.E319K Chr.1: 214332035-214332035 on Build GRCh38
6414691 c.967C>A p.L323I Chr.1: 214332047-214332047 on Build GRCh38
903696 c.957G>A p.E319E Chr.1: 214332037-214332037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377480785 Chr.1: 214332199-214332199 on Build GRCh38
rs748175763 Chr.1: 214332054-214332054 on Build GRCh38
rs973721636 Chr.1: 214332069-214332069 on Build GRCh38
rs769275348 Chr.1: 214332044-214332044 on Build GRCh38
rs763516758 Chr.1: 214332014-214332015 on Build GRCh38
rs751122379 Chr.1: 214332152-214332152 on Build GRCh38
rs766661501 Chr.1: 214332065-214332065 on Build GRCh38
rs370210032 Chr.1: 214332025-214332025 on Build GRCh38
rs368794892 Chr.1: 214332187-214332187 on Build GRCh38
rs963873702 Chr.1: 214332060-214332060 on Build GRCh38
rs35289970 Chr.1: 214332024-214332024 on Build GRCh38
rs535733694 Chr.1: 214332155-214332155 on Build GRCh38
rs200825251 Chr.1: 214332126-214332126 on Build GRCh38
rs772296667 Chr.1: 214332055-214332055 on Build GRCh38
rs779509405 Chr.1: 214332175-214332175 on Build GRCh38
rs559965377 Chr.1: 214332169-214332169 on Build GRCh38
rs755402563 Chr.1: 214332168-214332168 on Build GRCh38
rs1055699705 Chr.1: 214332045-214332045 on Build GRCh38
rs757855784 Chr.1: 214332105-214332105 on Build GRCh38
rs376272776 Chr.1: 214332085-214332085 on Build GRCh38
rs141785703 Chr.1: 214332088-214332088 on Build GRCh38
rs779044545 Chr.1: 214332192-214332192 on Build GRCh38
rs766703409 Chr.1: 214332122-214332122 on Build GRCh38
rs201487916 Chr.1: 214332020-214332020 on Build GRCh38
rs780252872 Chr.1: 214332024-214332024 on Build GRCh38
rs138541019 Chr.1: 214332157-214332157 on Build GRCh38
rs867430691 Chr.1: 214332023-214332023 on Build GRCh38
rs752635538 Chr.1: 214332096-214332096 on Build GRCh38
rs780958680 Chr.1: 214332158-214332158 on Build GRCh38
rs1052794443 Chr.1: 214332166-214332166 on Build GRCh38
rs776266188 Chr.1: 214332066-214332066 on Build GRCh38
rs773431617 Chr.1: 214332061-214332061 on Build GRCh38
rs765154080 Chr.1: 214332089-214332089 on Build GRCh38
rs749669555 Chr.1: 214332165-214332165 on Build GRCh38
rs760834330 Chr.1: 214332063-214332063 on Build GRCh38
rs369157821 Chr.1: 214332067-214332067 on Build GRCh38

View Full Product Details