Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28996039-28996264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF311
Entrez Gene ID
282890
Gene Name
zinc finger protein 311
Gene Aliases
zf31
UniGene
Hs.148218
Gene Chromosome Location
Chr.6: 28994785-29005668 on Build GRCh38
Transcripts
NM_001010877.3 NM_001318534.1 NM_001318535.1 XM_005249019.3 XM_006715067.2 XM_011514510.1 XM_011514511.2 XM_011514512.1 XM_011514513.1 XM_011514514.1 XM_011514516.1 XM_011514517.1 XM_011514518.1 XM_017010756.1 XM_017010757.1 XM_017010758.1 XM_017010759.1 XM_017010760.1 XM_017010761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
741596 c.743A>C p.E248A Chr.6: 28996259-28996259 on Build GRCh38
5569734 c.916A>G p.T306A Chr.6: 28996086-28996086 on Build GRCh38
184728 c.790C>T p.L264F Chr.6: 28996212-28996212 on Build GRCh38
1077186 c.886A>G p.I296V Chr.6: 28996116-28996116 on Build GRCh38
741597 c.856A>G p.T286A Chr.6: 28996146-28996146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770152008 Chr.6: 28996194-28996194 on Build GRCh38
rs377545660 Chr.6: 28996089-28996089 on Build GRCh38
rs780836230 Chr.6: 28996169-28996169 on Build GRCh38
rs745508403 Chr.6: 28996182-28996182 on Build GRCh38
rs754567154 Chr.6: 28996134-28996134 on Build GRCh38
rs1029506310 Chr.6: 28996243-28996243 on Build GRCh38
rs1014149924 Chr.6: 28996062-28996062 on Build GRCh38
rs758386496 Chr.6: 28996097-28996097 on Build GRCh38
rs964013405 Chr.6: 28996153-28996153 on Build GRCh38
rs756884037 Chr.6: 28996155-28996155 on Build GRCh38
rs752632862 Chr.6: 28996096-28996096 on Build GRCh38
rs769151142 Chr.6: 28996226-28996226 on Build GRCh38
rs754977321 Chr.6: 28996067-28996067 on Build GRCh38
rs370813308 Chr.6: 28996232-28996232 on Build GRCh38
rs1007547246 Chr.6: 28996116-28996116 on Build GRCh38
rs569044558 Chr.6: 28996064-28996064 on Build GRCh38
rs749697633 Chr.6: 28996208-28996208 on Build GRCh38
rs778940570 Chr.6: 28996091-28996091 on Build GRCh38
rs1022376699 Chr.6: 28996083-28996083 on Build GRCh38
rs548059546 Chr.6: 28996159-28996159 on Build GRCh38
rs780300338 Chr.6: 28996203-28996203 on Build GRCh38
rs751107430 Chr.6: 28996122-28996122 on Build GRCh38
rs748265670 Chr.6: 28996107-28996108 on Build GRCh38

View Full Product Details