Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 5172019-5172280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL8B
Entrez Gene ID
55207
Gene Name
ADP ribosylation factor like GTPase 8B
Gene Aliases
ARL10C, Gie1
UniGene
Hs.250009
Gene Chromosome Location
Chr.3: 5122245-5180916 on Build GRCh38
Transcripts
NM_018184.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6010782 c.235C>T p.R79* Chr.3: 5172180-5172180 on Build GRCh38
2852544 c.266T>C p.V89A Chr.3: 5172211-5172211 on Build GRCh38
7135636 c.230G>T p.R77L Chr.3: 5172175-5172175 on Build GRCh38
7135635 c.230G>T p.R77L Chr.3: 5172175-5172175 on Build GRCh38
6255502 c.257G>T p.C86F Chr.3: 5172202-5172202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777576635 Chr.3: 5172101-5172101 on Build GRCh38
rs201621702 Chr.3: 5172173-5172173 on Build GRCh38
rs749158799 Chr.3: 5172102-5172102 on Build GRCh38
rs775906506 Chr.3: 5172152-5172152 on Build GRCh38
rs767398685 Chr.3: 5172130-5172131 on Build GRCh38
rs73002996 Chr.3: 5172137-5172137 on Build GRCh38
rs531664639 Chr.3: 5172197-5172197 on Build GRCh38
rs201226709 Chr.3: 5172145-5172145 on Build GRCh38
rs907265533 Chr.3: 5172092-5172092 on Build GRCh38
rs991384147 Chr.3: 5172226-5172226 on Build GRCh38
rs767052632 Chr.3: 5172219-5172219 on Build GRCh38
rs1007116032 Chr.3: 5172063-5172063 on Build GRCh38
rs949498357 Chr.3: 5172232-5172232 on Build GRCh38
rs755951287 Chr.3: 5172099-5172099 on Build GRCh38
rs371474083 Chr.3: 5172116-5172116 on Build GRCh38
rs761002964 Chr.3: 5172158-5172158 on Build GRCh38
rs777138304 Chr.3: 5172174-5172174 on Build GRCh38
rs752296356 Chr.3: 5172231-5172231 on Build GRCh38
rs202026045 Chr.3: 5172164-5172164 on Build GRCh38
rs867184471 Chr.3: 5172065-5172065 on Build GRCh38
rs755964598 Chr.3: 5172231-5172232 on Build GRCh38
rs753692125 Chr.3: 5172245-5172245 on Build GRCh38
rs200689326 Chr.3: 5172243-5172243 on Build GRCh38
rs889726955 Chr.3: 5172062-5172062 on Build GRCh38
rs759125519 Chr.3: 5172216-5172216 on Build GRCh38
rs779054052 Chr.3: 5172238-5172241 on Build GRCh38
rs1019470772 Chr.3: 5172064-5172064 on Build GRCh38
rs552473513 Chr.3: 5172232-5172232 on Build GRCh38
rs373896889 Chr.3: 5172066-5172066 on Build GRCh38
rs1045856626 Chr.3: 5172067-5172067 on Build GRCh38
rs983803728 Chr.3: 5172141-5172141 on Build GRCh38
rs1025399433 Chr.3: 5172110-5172110 on Build GRCh38
rs762347837 Chr.3: 5172194-5172194 on Build GRCh38
rs777488623 Chr.3: 5172242-5172242 on Build GRCh38
rs751027401 Chr.3: 5172201-5172201 on Build GRCh38
rs755769192 Chr.3: 5172237-5172237 on Build GRCh38
rs113873889 Chr.3: 5172043-5172043 on Build GRCh38
rs770914636 Chr.3: 5172115-5172115 on Build GRCh38
rs978108116 Chr.3: 5172235-5172235 on Build GRCh38
rs201567012 Chr.3: 5172195-5172195 on Build GRCh38

View Full Product Details