Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42202223-42202480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAT5B
Entrez Gene ID
6777
Gene Name
signal transducer and activator of transcription 5B
Gene Aliases
STAT5
UniGene
Hs.595276
Gene Chromosome Location
Chr.17: 42199177-42276460 on Build GRCh38
Transcripts
NM_012448.3 XM_017024977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6009009 c.2144C>T p.S715F Chr.17: 42202433-42202433 on Build GRCh38
1710302 c.2206C>T p.P736S Chr.17: 42202371-42202371 on Build GRCh38
979451 c.2140G>A p.A714T Chr.17: 42202437-42202437 on Build GRCh38
6791713 c.2159delG p.G720fs*46 Chr.17: 42202418-42202418 on Build GRCh38
6929068 c.2188C>T p.P730S Chr.17: 42202389-42202389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752593400 Chr.17: 42202437-42202437 on Build GRCh38
rs768971207 Chr.17: 42202399-42202399 on Build GRCh38
rs760954156 Chr.17: 42202268-42202268 on Build GRCh38
rs767216293 Chr.17: 42202411-42202411 on Build GRCh38
rs771324059 Chr.17: 42202324-42202324 on Build GRCh38
rs1030951245 Chr.17: 42202272-42202272 on Build GRCh38
rs769715895 Chr.17: 42202460-42202460 on Build GRCh38
rs1047863698 Chr.17: 42202316-42202316 on Build GRCh38
rs1000786167 Chr.17: 42202269-42202269 on Build GRCh38
rs769087102 Chr.17: 42202302-42202302 on Build GRCh38
rs747200583 Chr.17: 42202315-42202315 on Build GRCh38
rs200042237 Chr.17: 42202392-42202392 on Build GRCh38
rs368370171 Chr.17: 42202266-42202266 on Build GRCh38
rs1056739990 Chr.17: 42202246-42202246 on Build GRCh38
rs896307409 Chr.17: 42202249-42202249 on Build GRCh38
rs529306345 Chr.17: 42202417-42202417 on Build GRCh38
rs746119686 Chr.17: 42202450-42202450 on Build GRCh38
rs777142714 Chr.17: 42202318-42202318 on Build GRCh38
rs1037612150 Chr.17: 42202277-42202277 on Build GRCh38
rs781501709 Chr.17: 42202447-42202447 on Build GRCh38
rs113762956 Chr.17: 42202325-42202325 on Build GRCh38
rs1045132966 Chr.17: 42202455-42202455 on Build GRCh38
rs867806419 Chr.17: 42202288-42202288 on Build GRCh38
rs750095303 Chr.17: 42202416-42202416 on Build GRCh38
rs754679455 Chr.17: 42202308-42202308 on Build GRCh38
rs139131471 Chr.17: 42202406-42202406 on Build GRCh38
rs372734271 Chr.17: 42202362-42202362 on Build GRCh38
rs769925436 Chr.17: 42202353-42202353 on Build GRCh38
rs774739413 Chr.17: 42202405-42202405 on Build GRCh38
rs754980286 Chr.17: 42202429-42202429 on Build GRCh38
rs957214879 Chr.17: 42202273-42202273 on Build GRCh38
rs1012758549 Chr.17: 42202261-42202261 on Build GRCh38
rs779789198 Chr.17: 42202299-42202299 on Build GRCh38
rs985226739 Chr.17: 42202349-42202349 on Build GRCh38
rs756064597 Chr.17: 42202293-42202293 on Build GRCh38
rs941014817 Chr.17: 42202377-42202377 on Build GRCh38
rs929910807 Chr.17: 42202410-42202410 on Build GRCh38
rs745998088 Chr.17: 42202347-42202347 on Build GRCh38
rs551009906 Chr.17: 42202425-42202425 on Build GRCh38
rs749983615 Chr.17: 42202289-42202289 on Build GRCh38
rs201825895 Chr.17: 42202438-42202438 on Build GRCh38
rs374819724 Chr.17: 42202309-42202309 on Build GRCh38
rs143157773 Chr.17: 42202423-42202423 on Build GRCh38
rs778825869 Chr.17: 42202435-42202435 on Build GRCh38
rs753827534 Chr.17: 42202305-42202305 on Build GRCh38

View Full Product Details