Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35421866-35422137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLFN12
Entrez Gene ID
55106
Gene Name
schlafen family member 12
Gene Aliases
SLFN3
UniGene
Hs.663548
Gene Chromosome Location
Chr.17: 35406684-35433176 on Build GRCh38
Transcripts
NM_001289009.1 NM_018042.4 XM_005257995.4 XM_011524967.2 XM_017024809.1 XM_017024810.1 XM_017024811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1710152 c.1014C>G p.I338M Chr.17: 35422015-35422015 on Build GRCh38
977920 c.926G>A p.R309H Chr.17: 35422103-35422103 on Build GRCh38
977919 c.1005G>T p.K335N Chr.17: 35422024-35422024 on Build GRCh38
1640649 c.1000A>C p.R334R Chr.17: 35422029-35422029 on Build GRCh38
5891517 c.976G>A p.D326N Chr.17: 35422053-35422053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946735541 Chr.17: 35422012-35422012 on Build GRCh38
rs754307307 Chr.17: 35422028-35422028 on Build GRCh38
rs766812481 Chr.17: 35422026-35422026 on Build GRCh38
rs895676385 Chr.17: 35422074-35422074 on Build GRCh38
rs968551105 Chr.17: 35421937-35421937 on Build GRCh38
rs958559405 Chr.17: 35422009-35422009 on Build GRCh38
rs917174039 Chr.17: 35422046-35422046 on Build GRCh38
rs1042912923 Chr.17: 35422015-35422015 on Build GRCh38
rs2671817 Chr.17: 35421920-35421920 on Build GRCh38
rs117621072 Chr.17: 35421981-35421981 on Build GRCh38
rs369876826 Chr.17: 35421976-35421976 on Build GRCh38
rs755870788 Chr.17: 35422051-35422051 on Build GRCh38
rs773780103 Chr.17: 35422008-35422008 on Build GRCh38
rs747656589 Chr.17: 35422093-35422093 on Build GRCh38
rs920865334 Chr.17: 35421908-35421908 on Build GRCh38
rs771820408 Chr.17: 35421988-35421988 on Build GRCh38
rs758593515 Chr.17: 35421967-35421967 on Build GRCh38
rs771912550 Chr.17: 35422103-35422103 on Build GRCh38
rs113965176 Chr.17: 35421906-35421906 on Build GRCh38
rs752739868 Chr.17: 35422036-35422036 on Build GRCh38
rs386796756 Chr.17: 35421959-35421960 on Build GRCh38
rs552992473 Chr.17: 35422108-35422108 on Build GRCh38
rs772945700 Chr.17: 35422104-35422104 on Build GRCh38
rs761111197 Chr.17: 35422024-35422024 on Build GRCh38
rs372943525 Chr.17: 35421984-35421984 on Build GRCh38
rs761129234 Chr.17: 35422107-35422107 on Build GRCh38
rs375512152 Chr.17: 35421944-35421944 on Build GRCh38
rs778607281 Chr.17: 35422092-35422092 on Build GRCh38
rs185599318 Chr.17: 35422109-35422109 on Build GRCh38
rs759568022 Chr.17: 35422031-35422031 on Build GRCh38
rs765404619 Chr.17: 35422035-35422035 on Build GRCh38
rs749198608 Chr.17: 35422077-35422077 on Build GRCh38
rs376178511 Chr.17: 35422090-35422090 on Build GRCh38
rs953486795 Chr.17: 35421915-35421915 on Build GRCh38
rs758683251 Chr.17: 35422037-35422037 on Build GRCh38
rs973622685 Chr.17: 35421897-35421897 on Build GRCh38
rs769677899 Chr.17: 35421972-35421972 on Build GRCh38
rs779885382 Chr.17: 35422054-35422054 on Build GRCh38
rs904455026 Chr.17: 35422030-35422030 on Build GRCh38
rs748751459 Chr.17: 35421980-35421980 on Build GRCh38
rs537149546 Chr.17: 35421918-35421918 on Build GRCh38
rs1056034033 Chr.17: 35422060-35422060 on Build GRCh38
rs781002150 Chr.17: 35421971-35421971 on Build GRCh38
rs777954946 Chr.17: 35422040-35422040 on Build GRCh38
rs750231522 Chr.17: 35422047-35422047 on Build GRCh38

View Full Product Details