Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 48761265-48761538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTIF
Entrez Gene ID
9811
Gene Name
cap binding complex dependent translation initiation factor
Gene Aliases
Gm672, KIAA0427
UniGene
Hs.145230
Gene Chromosome Location
Chr.18: 48539056-48863217 on Build GRCh38
Transcripts
NM_001142397.1 NM_014772.2 XM_005258392.4 XM_006722583.3 XM_006722586.3 XM_006722587.3 XM_006722588.3 XM_006722589.3 XM_011526278.2 XM_011526279.2 XM_017026100.1 XM_017026101.1 XM_017026102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
116316 c.1107G>C p.Q369H Chr.18: 48761425-48761425 on Build GRCh38
6637464 c.932A>G p.E311G Chr.18: 48761394-48761394 on Build GRCh38
5750261 c.1021G>A p.V341M Chr.18: 48761483-48761483 on Build GRCh38
4072394 c.1194G>T p.E398D Chr.18: 48761512-48761512 on Build GRCh38
988724 c.940G>A p.V314M Chr.18: 48761402-48761402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770680402 Chr.18: 48761372-48761372 on Build GRCh38
rs775288854 Chr.18: 48761446-48761446 on Build GRCh38
rs759965898 Chr.18: 48761474-48761474 on Build GRCh38
rs754211947 Chr.18: 48761397-48761397 on Build GRCh38
rs761006728 Chr.18: 48761466-48761466 on Build GRCh38
rs377077939 Chr.18: 48761386-48761386 on Build GRCh38
rs910330334 Chr.18: 48761337-48761337 on Build GRCh38
rs746872354 Chr.18: 48761365-48761365 on Build GRCh38
rs773375297 Chr.18: 48761465-48761465 on Build GRCh38
rs751006365 Chr.18: 48761483-48761483 on Build GRCh38
rs141427281 Chr.18: 48761410-48761410 on Build GRCh38
rs745605627 Chr.18: 48761440-48761440 on Build GRCh38
rs769833122 Chr.18: 48761442-48761442 on Build GRCh38
rs182469390 Chr.18: 48761387-48761387 on Build GRCh38
rs761405553 Chr.18: 48761369-48761372 on Build GRCh38
rs994309612 Chr.18: 48761285-48761285 on Build GRCh38
rs117160953 Chr.18: 48761377-48761377 on Build GRCh38
rs568317476 Chr.18: 48761302-48761302 on Build GRCh38
rs747541350 Chr.18: 48761339-48761339 on Build GRCh38
rs187024603 Chr.18: 48761482-48761482 on Build GRCh38
rs756915974 Chr.18: 48761506-48761506 on Build GRCh38
rs370440560 Chr.18: 48761441-48761441 on Build GRCh38
rs202183503 Chr.18: 48761402-48761402 on Build GRCh38
rs946114862 Chr.18: 48761439-48761439 on Build GRCh38
rs780971462 Chr.18: 48761427-48761427 on Build GRCh38
rs772725922 Chr.18: 48761349-48761349 on Build GRCh38
rs755542623 Chr.18: 48761401-48761401 on Build GRCh38
rs745732684 Chr.18: 48761517-48761517 on Build GRCh38
rs772025886 Chr.18: 48761453-48761453 on Build GRCh38
rs757973945 Chr.18: 48761480-48761480 on Build GRCh38
rs753112740 Chr.18: 48761479-48761479 on Build GRCh38
rs553095841 Chr.18: 48761447-48761447 on Build GRCh38
rs550186960 Chr.18: 48761331-48761331 on Build GRCh38
rs138506490 Chr.18: 48761290-48761290 on Build GRCh38
rs770531562 Chr.18: 48761415-48761415 on Build GRCh38
rs746688468 Chr.18: 48761413-48761413 on Build GRCh38
rs774507022 Chr.18: 48761384-48761384 on Build GRCh38
rs1026241661 Chr.18: 48761307-48761307 on Build GRCh38
rs980392292 Chr.18: 48761403-48761403 on Build GRCh38
rs767134546 Chr.18: 48761407-48761409 on Build GRCh38
rs371941280 Chr.18: 48761385-48761385 on Build GRCh38
rs375114552 Chr.18: 48761379-48761379 on Build GRCh38
rs989175974 Chr.18: 48761361-48761361 on Build GRCh38
rs952965041 Chr.18: 48761336-48761336 on Build GRCh38
rs771769098 Chr.18: 48761348-48761348 on Build GRCh38
rs759502704 Chr.18: 48761378-48761378 on Build GRCh38
rs765838377 Chr.18: 48761475-48761475 on Build GRCh38
rs754263370 Chr.18: 48761467-48761467 on Build GRCh38

View Full Product Details