Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64284712-64284951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR137
Entrez Gene ID
56834
Gene Name
G protein-coupled receptor 137
Gene Aliases
C11orf4, GPR137A, TM7SF1L1
UniGene
Hs.523763
Gene Chromosome Location
Chr.11: 64270337-64289500 on Build GRCh38
Transcripts
NM_001170726.1 NM_001170881.1 NM_001177358.1 XM_005274101.2 XM_011545168.2 XM_011545170.2 XM_011545171.2 XM_011545172.2 XM_017018014.1 XM_017018016.1

SNP Detail

SNP ID Chromosome Location
rs375058825 Chr.11: 64284769-64284769 on Build GRCh38
rs772761198 Chr.11: 64284772-64284772 on Build GRCh38
rs1027982025 Chr.11: 64284766-64284766 on Build GRCh38
rs889834107 Chr.11: 64284808-64284808 on Build GRCh38
rs559870176 Chr.11: 64284889-64284889 on Build GRCh38
rs652611 Chr.11: 64284834-64284834 on Build GRCh38
rs944798802 Chr.11: 64284846-64284846 on Build GRCh38
rs563409227 Chr.11: 64284869-64284869 on Build GRCh38
rs1040953380 Chr.11: 64284731-64284731 on Build GRCh38
rs763067502 Chr.11: 64284800-64284801 on Build GRCh38
rs932178667 Chr.11: 64284866-64284866 on Build GRCh38
rs543422577 Chr.11: 64284856-64284856 on Build GRCh38
rs541370892 Chr.11: 64284811-64284811 on Build GRCh38
rs979318645 Chr.11: 64284826-64284826 on Build GRCh38
rs773441698 Chr.11: 64284782-64284782 on Build GRCh38
rs995487552 Chr.11: 64284795-64284795 on Build GRCh38
rs552223505 Chr.11: 64284908-64284908 on Build GRCh38
rs920518344 Chr.11: 64284833-64284833 on Build GRCh38
rs532262628 Chr.11: 64284890-64284890 on Build GRCh38
rs902252804 Chr.11: 64284927-64284927 on Build GRCh38
rs1010499352 Chr.11: 64284918-64284918 on Build GRCh38
rs748639671 Chr.11: 64284771-64284771 on Build GRCh38
rs900737074 Chr.11: 64284737-64284737 on Build GRCh38
rs981954375 Chr.11: 64284802-64284802 on Build GRCh38
rs5031004 Chr.11: 64284836-64284836 on Build GRCh38
rs779169430 Chr.11: 64284761-64284761 on Build GRCh38
rs957668799 Chr.11: 64284768-64284768 on Build GRCh38
rs572555375 Chr.11: 64284801-64284801 on Build GRCh38
rs1019688224 Chr.11: 64284893-64284893 on Build GRCh38
rs752859657 Chr.11: 64284750-64284762 on Build GRCh38
rs1010531924 Chr.11: 64284770-64284770 on Build GRCh38
rs111793546 Chr.11: 64284800-64284800 on Build GRCh38
rs1020607088 Chr.11: 64284925-64284925 on Build GRCh38
rs927810128 Chr.11: 64284805-64284805 on Build GRCh38
rs968884764 Chr.11: 64284820-64284820 on Build GRCh38
rs917295477 Chr.11: 64284880-64284880 on Build GRCh38
rs991177813 Chr.11: 64284817-64284817 on Build GRCh38
rs1044528815 Chr.11: 64284910-64284910 on Build GRCh38

View Full Product Details