Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 111692369-111692594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
-
UniGene
Hs.428209
Gene Chromosome Location
Chr.9: 111686175-111795949 on Build GRCh38
Transcripts
NM_001080551.2 NM_173521.4 NR_109816.1 XM_006716974.2 XM_006716975.2 XM_011518302.2 XM_011518303.1 XM_011518306.1 XM_017014340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7298024 c.3390T>C p.S1130S Chr.9: 111692395-111692395 on Build GRCh38
7298025 c.3273T>C p.S1091S Chr.9: 111692395-111692395 on Build GRCh38
7283561 c.3407A>G p.D1136G Chr.9: 111692378-111692378 on Build GRCh38
7283562 c.3290A>G p.D1097G Chr.9: 111692378-111692378 on Build GRCh38
6616995 c.3364C>A p.Q1122K Chr.9: 111692421-111692421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758002551 Chr.9: 111692540-111692540 on Build GRCh38
rs779392476 Chr.9: 111692550-111692555 on Build GRCh38
rs916340157 Chr.9: 111692548-111692548 on Build GRCh38
rs200568507 Chr.9: 111692443-111692443 on Build GRCh38
rs765889023 Chr.9: 111692557-111692557 on Build GRCh38
rs762156009 Chr.9: 111692471-111692472 on Build GRCh38
rs750543623 Chr.9: 111692488-111692489 on Build GRCh38
rs537629803 Chr.9: 111692480-111692480 on Build GRCh38
rs764562944 Chr.9: 111692439-111692439 on Build GRCh38
rs754401106 Chr.9: 111692514-111692514 on Build GRCh38
rs751679362 Chr.9: 111692471-111692484 on Build GRCh38
rs568747748 Chr.9: 111692462-111692462 on Build GRCh38
rs186931900 Chr.9: 111692436-111692436 on Build GRCh38
rs939069286 Chr.9: 111692501-111692501 on Build GRCh38
rs746023441 Chr.9: 111692472-111692472 on Build GRCh38
rs750041009 Chr.9: 111692531-111692531 on Build GRCh38
rs777179619 Chr.9: 111692518-111692518 on Build GRCh38
rs774422432 Chr.9: 111692471-111692471 on Build GRCh38
rs765001734 Chr.9: 111692528-111692528 on Build GRCh38
rs761282811 Chr.9: 111692489-111692489 on Build GRCh38
rs756593299 Chr.9: 111692490-111692491 on Build GRCh38
rs755238768 Chr.9: 111692545-111692546 on Build GRCh38
rs762065327 Chr.9: 111692520-111692520 on Build GRCh38
rs780491032 Chr.9: 111692511-111692512 on Build GRCh38
rs772046616 Chr.9: 111692477-111692477 on Build GRCh38
rs989323279 Chr.9: 111692420-111692420 on Build GRCh38
rs746953682 Chr.9: 111692453-111692453 on Build GRCh38
rs546584336 Chr.9: 111692444-111692444 on Build GRCh38
rs767582583 Chr.9: 111692487-111692488 on Build GRCh38
rs148430343 Chr.9: 111692441-111692441 on Build GRCh38
rs12343237 Chr.9: 111692452-111692452 on Build GRCh38
rs73656294 Chr.9: 111692455-111692455 on Build GRCh38
rs771231552 Chr.9: 111692466-111692466 on Build GRCh38
rs753249231 Chr.9: 111692408-111692408 on Build GRCh38
rs554482408 Chr.9: 111692497-111692497 on Build GRCh38
rs751022884 Chr.9: 111692558-111692558 on Build GRCh38

View Full Product Details