Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 223749012-223749237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN2
Entrez Gene ID
824
Gene Name
calpain 2
Gene Aliases
CANP2, CANPL2, CANPml, mCANP
UniGene
Hs.350899
Gene Chromosome Location
Chr.1: 223701593-223776018 on Build GRCh38
Transcripts
NM_001146068.1 NM_001748.4 XM_011544286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1960501 c.787G>A p.A263T Chr.1: 223749096-223749096 on Build GRCh38
6619820 c.579G>T p.E193D Chr.1: 223749122-223749122 on Build GRCh38
6619813 c.553G>A p.A185T Chr.1: 223749096-223749096 on Build GRCh38
6619819 c.813G>T p.E271D Chr.1: 223749122-223749122 on Build GRCh38
3689373 c.808G>A p.E270K Chr.1: 223749117-223749117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773972885 Chr.1: 223749133-223749133 on Build GRCh38
rs777705012 Chr.1: 223749104-223749104 on Build GRCh38
rs984215768 Chr.1: 223749050-223749050 on Build GRCh38
rs754029627 Chr.1: 223749056-223749056 on Build GRCh38
rs773673860 Chr.1: 223749124-223749125 on Build GRCh38
rs757037832 Chr.1: 223749128-223749128 on Build GRCh38
rs371937820 Chr.1: 223749041-223749041 on Build GRCh38
rs992765811 Chr.1: 223749117-223749117 on Build GRCh38
rs10915715 Chr.1: 223749162-223749162 on Build GRCh38
rs535145373 Chr.1: 223749208-223749208 on Build GRCh38
rs908671232 Chr.1: 223749113-223749113 on Build GRCh38
rs149026969 Chr.1: 223749048-223749048 on Build GRCh38
rs376570741 Chr.1: 223749131-223749131 on Build GRCh38
rs755278339 Chr.1: 223749062-223749062 on Build GRCh38
rs752842266 Chr.1: 223749082-223749082 on Build GRCh38
rs746743653 Chr.1: 223749123-223749123 on Build GRCh38
rs769596501 Chr.1: 223749084-223749087 on Build GRCh38
rs927399955 Chr.1: 223749058-223749058 on Build GRCh38
rs761080563 Chr.1: 223749155-223749155 on Build GRCh38
rs373928571 Chr.1: 223749149-223749149 on Build GRCh38
rs765201577 Chr.1: 223749074-223749074 on Build GRCh38
rs958681867 Chr.1: 223749078-223749078 on Build GRCh38
rs777094351 Chr.1: 223749167-223749167 on Build GRCh38
rs758321767 Chr.1: 223749091-223749091 on Build GRCh38
rs889555861 Chr.1: 223749207-223749207 on Build GRCh38
rs773696661 Chr.1: 223749040-223749040 on Build GRCh38
rs772648739 Chr.1: 223749148-223749148 on Build GRCh38
rs570841429 Chr.1: 223749203-223749203 on Build GRCh38
rs982987003 Chr.1: 223749085-223749085 on Build GRCh38
rs1053527214 Chr.1: 223749164-223749164 on Build GRCh38
rs768523941 Chr.1: 223749132-223749132 on Build GRCh38
rs944278433 Chr.1: 223749127-223749127 on Build GRCh38
rs768202916 Chr.1: 223749031-223749031 on Build GRCh38
rs748864188 Chr.1: 223749147-223749147 on Build GRCh38
rs530919451 Chr.1: 223749101-223749101 on Build GRCh38
rs749217943 Chr.1: 223749159-223749159 on Build GRCh38
rs761133559 Chr.1: 223749047-223749047 on Build GRCh38

View Full Product Details