Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 84152649-84152878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP162
Entrez Gene ID
22832
Gene Name
centrosomal protein 162
Gene Aliases
C6orf84, KIAA1009, QN1
UniGene
Hs.485865
Gene Chromosome Location
Chr.6: 84124241-84227889 on Build GRCh38
Transcripts
NM_001286206.1 NM_014895.3 XM_005248674.3 XM_006715380.2 XM_011535592.2 XM_017010483.1 XM_017010484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4635439 c.3336G>A p.Q1112Q Chr.6: 84152838-84152838 on Build GRCh38
4628556 c.3428G>T p.S1143I Chr.6: 84152746-84152746 on Build GRCh38
5634395 c.3296G>C p.R1099T Chr.6: 84152878-84152878 on Build GRCh38
6020416 c.3401G>T p.R1134M Chr.6: 84152773-84152773 on Build GRCh38
1285741 c.3333T>A p.L1111L Chr.6: 84152841-84152841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962750553 Chr.6: 84152715-84152715 on Build GRCh38
rs1023307293 Chr.6: 84152838-84152838 on Build GRCh38
rs769554715 Chr.6: 84152679-84152679 on Build GRCh38
rs528479083 Chr.6: 84152784-84152784 on Build GRCh38
rs138633533 Chr.6: 84152818-84152818 on Build GRCh38
rs201802888 Chr.6: 84152713-84152713 on Build GRCh38
rs780046675 Chr.6: 84152760-84152760 on Build GRCh38
rs761309157 Chr.6: 84152711-84152711 on Build GRCh38
rs773670365 Chr.6: 84152692-84152692 on Build GRCh38
rs759084178 Chr.6: 84152693-84152693 on Build GRCh38
rs992381711 Chr.6: 84152739-84152739 on Build GRCh38
rs774043726 Chr.6: 84152793-84152793 on Build GRCh38
rs773029329 Chr.6: 84152682-84152682 on Build GRCh38
rs897597043 Chr.6: 84152763-84152763 on Build GRCh38
rs374681698 Chr.6: 84152790-84152790 on Build GRCh38
rs754273399 Chr.6: 84152726-84152726 on Build GRCh38
rs974242418 Chr.6: 84152724-84152724 on Build GRCh38
rs144773801 Chr.6: 84152780-84152780 on Build GRCh38
rs748086370 Chr.6: 84152776-84152776 on Build GRCh38
rs942198959 Chr.6: 84152718-84152718 on Build GRCh38
rs745559104 Chr.6: 84152796-84152796 on Build GRCh38
rs200738473 Chr.6: 84152785-84152785 on Build GRCh38
rs770584366 Chr.6: 84152689-84152689 on Build GRCh38
rs761211213 Chr.6: 84152815-84152815 on Build GRCh38
rs750634217 Chr.6: 84152823-84152823 on Build GRCh38
rs765576862 Chr.6: 84152821-84152821 on Build GRCh38
rs758489637 Chr.6: 84152753-84152753 on Build GRCh38
rs750676881 Chr.6: 84152743-84152743 on Build GRCh38
rs200917055 Chr.6: 84152792-84152792 on Build GRCh38
rs771584753 Chr.6: 84152797-84152797 on Build GRCh38
rs766997189 Chr.6: 84152694-84152694 on Build GRCh38
rs57366551 Chr.6: 84152827-84152827 on Build GRCh38
rs776044748 Chr.6: 84152709-84152709 on Build GRCh38
rs1037426220 Chr.6: 84152719-84152719 on Build GRCh38
rs774897791 Chr.6: 84152813-84152813 on Build GRCh38
rs765498400 Chr.6: 84152738-84152738 on Build GRCh38
rs372272481 Chr.6: 84152732-84152732 on Build GRCh38
rs764707783 Chr.6: 84152817-84152817 on Build GRCh38
rs748891806 Chr.6: 84152683-84152683 on Build GRCh38
rs895518334 Chr.6: 84152798-84152798 on Build GRCh38

View Full Product Details