Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129134001-129134260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISY1
Entrez Gene ID
57461
Gene Name
ISY1 splicing factor homolog
Gene Aliases
FSAP33
UniGene
Hs.512661
Gene Chromosome Location
Chr.3: 129127416-129161230 on Build GRCh38
Transcripts
NM_001199469.1 NM_020701.3 NM_001204890.1

Gene Symbol
ISY1-RAB43
Entrez Gene ID
100534599
Gene Name
ISY1-RAB43 readthrough
Gene Aliases
-
UniGene
Hs.512661
Gene Chromosome Location
Chr.3: 129087569-129161230 on Build GRCh38
Transcripts
NM_001199469.1 NM_020701.3 NM_001204890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6163262 c.662G>A p.R221K Chr.3: 129134141-129134141 on Build GRCh38
1418793 c.593C>A p.A198E Chr.3: 129134144-129134144 on Build GRCh38
728572 c.574G>A p.E192K Chr.3: 129134163-129134163 on Build GRCh38
4784807 c.659C>A p.A220E Chr.3: 129134144-129134144 on Build GRCh38
728573 c.586C>A p.R196R Chr.3: 129134151-129134151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765392126 Chr.3: 129134151-129134151 on Build GRCh38
rs772706716 Chr.3: 129134230-129134230 on Build GRCh38
rs760557661 Chr.3: 129134052-129134053 on Build GRCh38
rs188793366 Chr.3: 129134219-129134219 on Build GRCh38
rs769182087 Chr.3: 129134083-129134083 on Build GRCh38
rs772273195 Chr.3: 129134124-129134124 on Build GRCh38
rs763997533 Chr.3: 129134104-129134112 on Build GRCh38
rs919717438 Chr.3: 129134206-129134206 on Build GRCh38
rs758370541 Chr.3: 129134159-129134159 on Build GRCh38
rs79550573 Chr.3: 129134074-129134074 on Build GRCh38
rs752528385 Chr.3: 129134154-129134154 on Build GRCh38
rs908448973 Chr.3: 129134082-129134082 on Build GRCh38
rs779634710 Chr.3: 129134226-129134226 on Build GRCh38
rs369012886 Chr.3: 129134093-129134093 on Build GRCh38
rs777646912 Chr.3: 129134028-129134028 on Build GRCh38
rs745438915 Chr.3: 129134077-129134077 on Build GRCh38
rs758088143 Chr.3: 129134027-129134027 on Build GRCh38
rs745348660 Chr.3: 129134208-129134208 on Build GRCh38
rs910968961 Chr.3: 129134141-129134141 on Build GRCh38
rs780675003 Chr.3: 129134056-129134056 on Build GRCh38
rs753708427 Chr.3: 129134117-129134119 on Build GRCh38
rs746605008 Chr.3: 129134036-129134036 on Build GRCh38
rs748540779 Chr.3: 129134227-129134227 on Build GRCh38
rs573583959 Chr.3: 129134023-129134023 on Build GRCh38
rs986453015 Chr.3: 129134108-129134108 on Build GRCh38
rs753947979 Chr.3: 129134150-129134150 on Build GRCh38
rs144548237 Chr.3: 129134044-129134044 on Build GRCh38
rs762409591 Chr.3: 129134109-129134109 on Build GRCh38
rs372937929 Chr.3: 129134139-129134139 on Build GRCh38
rs773634203 Chr.3: 129134130-129134130 on Build GRCh38
rs201867968 Chr.3: 129134136-129134136 on Build GRCh38
rs563215458 Chr.3: 129134187-129134187 on Build GRCh38
rs376950781 Chr.3: 129134188-129134188 on Build GRCh38
rs777364741 Chr.3: 129134185-129134185 on Build GRCh38
rs766768318 Chr.3: 129134142-129134142 on Build GRCh38

View Full Product Details