Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 108798118-108798361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER
Entrez Gene ID
2241
Gene Name
FER tyrosine kinase
Gene Aliases
PPP1R74, TYK3, p94-Fer
UniGene
Hs.107418
Gene Chromosome Location
Chr.5: 108746085-109196841 on Build GRCh38
Transcripts
NM_001308028.1 NM_001308038.1 NM_005246.3 XM_011543266.1 XM_011543267.1 XM_011543269.1 XM_011543270.2 XM_011543271.2 XM_011543272.1 XM_011543278.1 XM_017009229.1 XM_017009230.1 XM_017009231.1 XM_017009232.1 XM_017009234.1 XM_017009235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
98896 c.169A>G p.S57G Chr.5: 108798351-108798351 on Build GRCh38
1059370 c.87G>T p.K29N Chr.5: 108798269-108798269 on Build GRCh38
6535654 c.54A>G p.Q18Q Chr.5: 108798236-108798236 on Build GRCh38
4769422 c.165G>A p.K55K Chr.5: 108798347-108798347 on Build GRCh38
6529572 c.151A>C p.N51H Chr.5: 108798333-108798333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998227821 Chr.5: 108798308-108798308 on Build GRCh38
rs781096339 Chr.5: 108798234-108798234 on Build GRCh38
rs141543178 Chr.5: 108798164-108798164 on Build GRCh38
rs900854674 Chr.5: 108798294-108798294 on Build GRCh38
rs575745920 Chr.5: 108798147-108798147 on Build GRCh38
rs776972232 Chr.5: 108798311-108798311 on Build GRCh38
rs779002488 Chr.5: 108798145-108798145 on Build GRCh38
rs934592965 Chr.5: 108798168-108798168 on Build GRCh38
rs771577555 Chr.5: 108798156-108798156 on Build GRCh38
rs925734590 Chr.5: 108798161-108798161 on Build GRCh38
rs35665762 Chr.5: 108798243-108798244 on Build GRCh38
rs747071972 Chr.5: 108798300-108798300 on Build GRCh38
rs147858956 Chr.5: 108798249-108798249 on Build GRCh38
rs1053290196 Chr.5: 108798175-108798175 on Build GRCh38
rs765674931 Chr.5: 108798173-108798173 on Build GRCh38
rs777332921 Chr.5: 108798160-108798160 on Build GRCh38
rs746165330 Chr.5: 108798313-108798313 on Build GRCh38
rs144530313 Chr.5: 108798239-108798239 on Build GRCh38
rs771199420 Chr.5: 108798301-108798301 on Build GRCh38
rs762689535 Chr.5: 108798214-108798214 on Build GRCh38
rs763556445 Chr.5: 108798221-108798221 on Build GRCh38
rs2229085 Chr.5: 108798266-108798266 on Build GRCh38
rs773453253 Chr.5: 108798149-108798149 on Build GRCh38
rs1049496902 Chr.5: 108798327-108798327 on Build GRCh38
rs751077094 Chr.5: 108798223-108798223 on Build GRCh38
rs757132456 Chr.5: 108798225-108798225 on Build GRCh38
rs758055737 Chr.5: 108798193-108798193 on Build GRCh38
rs752270983 Chr.5: 108798188-108798188 on Build GRCh38
rs142838306 Chr.5: 108798276-108798276 on Build GRCh38
rs761196807 Chr.5: 108798154-108798154 on Build GRCh38
rs760139281 Chr.5: 108798315-108798315 on Build GRCh38
rs770006209 Chr.5: 108798331-108798331 on Build GRCh38
rs777941768 Chr.5: 108798288-108798288 on Build GRCh38
rs199698325 Chr.5: 108798250-108798250 on Build GRCh38

View Full Product Details