Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168906659-168906884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC2
Entrez Gene ID
57818
Gene Name
glucose-6-phosphatase catalytic subunit 2
Gene Aliases
IGRP
UniGene
Hs.283963
Gene Chromosome Location
Chr.2: 168901223-168910000 on Build GRCh38
Transcripts
NM_001081686.1 NM_021176.2 XM_011511564.2 XM_011511565.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1400995 c.457C>A p.L153I Chr.2: 168906680-168906680 on Build GRCh38
4086538 c.489T>A p.S163R Chr.2: 168906712-168906712 on Build GRCh38
1325774 c.493T>C p.C165R Chr.2: 168906716-168906716 on Build GRCh38
5908102 c.544G>A p.G182R Chr.2: 168906767-168906767 on Build GRCh38
3314636 c.526C>T p.P176S Chr.2: 168906749-168906749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563271270 Chr.2: 168906786-168906786 on Build GRCh38
rs997529629 Chr.2: 168906687-168906687 on Build GRCh38
rs367627743 Chr.2: 168906804-168906804 on Build GRCh38
rs756644404 Chr.2: 168906785-168906785 on Build GRCh38
rs370478326 Chr.2: 168906793-168906793 on Build GRCh38
rs143071679 Chr.2: 168906858-168906858 on Build GRCh38
rs138726309 Chr.2: 168906752-168906752 on Build GRCh38
rs756521075 Chr.2: 168906692-168906692 on Build GRCh38
rs888480597 Chr.2: 168906719-168906719 on Build GRCh38
rs778517641 Chr.2: 168906730-168906730 on Build GRCh38
rs752937225 Chr.2: 168906776-168906776 on Build GRCh38
rs977773202 Chr.2: 168906805-168906805 on Build GRCh38
rs774625522 Chr.2: 168906763-168906763 on Build GRCh38
rs376500857 Chr.2: 168906755-168906755 on Build GRCh38
rs779907176 Chr.2: 168906803-168906803 on Build GRCh38
rs2232322 Chr.2: 168906734-168906734 on Build GRCh38
rs757934580 Chr.2: 168906798-168906798 on Build GRCh38
rs768651839 Chr.2: 168906745-168906745 on Build GRCh38
rs145050507 Chr.2: 168906735-168906735 on Build GRCh38
rs781242281 Chr.2: 168906820-168906820 on Build GRCh38
rs763091287 Chr.2: 168906809-168906809 on Build GRCh38
rs748755149 Chr.2: 168906699-168906699 on Build GRCh38
rs200099602 Chr.2: 168906797-168906797 on Build GRCh38
rs751915325 Chr.2: 168906684-168906684 on Build GRCh38
rs756923876 Chr.2: 168906727-168906727 on Build GRCh38
rs906548989 Chr.2: 168906834-168906834 on Build GRCh38
rs746984825 Chr.2: 168906743-168906743 on Build GRCh38
rs933654151 Chr.2: 168906739-168906739 on Build GRCh38
rs755335802 Chr.2: 168906686-168906686 on Build GRCh38
rs201647518 Chr.2: 168906824-168906824 on Build GRCh38
rs753230056 Chr.2: 168906691-168906691 on Build GRCh38
rs767972702 Chr.2: 168906688-168906688 on Build GRCh38
rs759872383 Chr.2: 168906766-168906766 on Build GRCh38
rs934799164 Chr.2: 168906844-168906844 on Build GRCh38
rs373009921 Chr.2: 168906787-168906787 on Build GRCh38
rs777268524 Chr.2: 168906697-168906697 on Build GRCh38
rs756731608 Chr.2: 168906791-168906791 on Build GRCh38
rs763080884 Chr.2: 168906753-168906753 on Build GRCh38
rs767933484 Chr.2: 168906774-168906774 on Build GRCh38
rs1049742604 Chr.2: 168906698-168906698 on Build GRCh38

View Full Product Details