Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 50535408-50535669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAF1
Entrez Gene ID
11124
Gene Name
Fas associated factor 1
Gene Aliases
CGI-03, HFAF1s, UBXD12, UBXN3A, hFAF1
UniGene
Hs.530402
Gene Chromosome Location
Chr.1: 50441263-50960264 on Build GRCh38
Transcripts
NM_007051.2 XM_017000136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738559 c.1420G>A p.D474N Chr.1: 50535443-50535443 on Build GRCh38
3400863 c.1406-2A>G p.? Chr.1: 50535459-50535459 on Build GRCh38
6126998 c.1415C>T p.T472I Chr.1: 50535448-50535448 on Build GRCh38
2192228 c.1432A>G p.M478V Chr.1: 50535431-50535431 on Build GRCh38
6126999 c.689C>T p.T230I Chr.1: 50535448-50535448 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs964428055 Chr.1: 50535601-50535601 on Build GRCh38
rs776496543 Chr.1: 50535501-50535501 on Build GRCh38
rs777588800 Chr.1: 50535451-50535451 on Build GRCh38
rs983714717 Chr.1: 50535568-50535568 on Build GRCh38
rs746770224 Chr.1: 50535486-50535486 on Build GRCh38
rs967667409 Chr.1: 50535637-50535637 on Build GRCh38
rs770880683 Chr.1: 50535498-50535498 on Build GRCh38
rs561577289 Chr.1: 50535596-50535596 on Build GRCh38
rs972790513 Chr.1: 50535488-50535488 on Build GRCh38
rs771386910 Chr.1: 50535466-50535467 on Build GRCh38
rs546902399 Chr.1: 50535627-50535627 on Build GRCh38
rs181763744 Chr.1: 50535603-50535603 on Build GRCh38
rs774678020 Chr.1: 50535510-50535513 on Build GRCh38
rs543298078 Chr.1: 50535545-50535545 on Build GRCh38

View Full Product Details