Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160282138-160282363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX19
Entrez Gene ID
5824
Gene Name
peroxisomal biogenesis factor 19
Gene Aliases
D1S2223E, HK33, PBD12A, PMP1, PMPI, PXF, PXMP1
UniGene
Hs.517232
Gene Chromosome Location
Chr.1: 160276809-160285151 on Build GRCh38
Transcripts
NM_001193644.1 NM_002857.3 NR_036492.1 NR_036493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880680 c.471G>A p.M157I Chr.1: 160282162-160282162 on Build GRCh38
1472865 c.22A>G p.K8E Chr.1: 160282170-160282170 on Build GRCh38
4024124 c.27C>A p.A9A Chr.1: 160282165-160282165 on Build GRCh38
322603 c.451G>T p.E151* Chr.1: 160282182-160282182 on Build GRCh38
4024123 c.468C>A p.A156A Chr.1: 160282165-160282165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770461028 Chr.1: 160282187-160282187 on Build GRCh38
rs762131917 Chr.1: 160282207-160282207 on Build GRCh38
rs755087556 Chr.1: 160282169-160282169 on Build GRCh38
rs778358584 Chr.1: 160282190-160282190 on Build GRCh38
rs377564891 Chr.1: 160282230-160282230 on Build GRCh38
rs771657561 Chr.1: 160282198-160282198 on Build GRCh38
rs775148442 Chr.1: 160282206-160282206 on Build GRCh38
rs751891878 Chr.1: 160282276-160282276 on Build GRCh38
rs1043071771 Chr.1: 160282182-160282182 on Build GRCh38
rs539610788 Chr.1: 160282223-160282223 on Build GRCh38
rs905834830 Chr.1: 160282191-160282191 on Build GRCh38
rs187570304 Chr.1: 160282320-160282320 on Build GRCh38
rs370448833 Chr.1: 160282179-160282179 on Build GRCh38
rs766423323 Chr.1: 160282158-160282158 on Build GRCh38
rs766912987 Chr.1: 160282222-160282222 on Build GRCh38
rs750013486 Chr.1: 160282195-160282195 on Build GRCh38
rs751797704 Chr.1: 160282159-160282159 on Build GRCh38
rs752489082 Chr.1: 160282176-160282176 on Build GRCh38
rs956441849 Chr.1: 160282340-160282340 on Build GRCh38
rs576265326 Chr.1: 160282268-160282268 on Build GRCh38
rs1029446738 Chr.1: 160282333-160282333 on Build GRCh38
rs183427802 Chr.1: 160282224-160282224 on Build GRCh38
rs756002277 Chr.1: 160282177-160282177 on Build GRCh38
rs575532165 Chr.1: 160282232-160282232 on Build GRCh38
rs928649830 Chr.1: 160282178-160282180 on Build GRCh38
rs41265791 Chr.1: 160282211-160282211 on Build GRCh38
rs770190209 Chr.1: 160282209-160282209 on Build GRCh38
rs753010045 Chr.1: 160282249-160282249 on Build GRCh38
rs745392627 Chr.1: 160282192-160282192 on Build GRCh38
rs749234062 Chr.1: 160282186-160282186 on Build GRCh38
rs1017941060 Chr.1: 160282263-160282263 on Build GRCh38
rs1005845614 Chr.1: 160282236-160282236 on Build GRCh38
rs78340311 Chr.1: 160282174-160282174 on Build GRCh38
rs866356222 Chr.1: 160282229-160282229 on Build GRCh38

View Full Product Details