Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 973004-973241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHN1
Entrez Gene ID
84069
Gene Name
pleckstrin homology domain containing N1
Gene Aliases
-
UniGene
Hs.528691
Gene Chromosome Location
Chr.1: 965820-975108 on Build GRCh38
Transcripts
NM_001160184.1 NM_032129.2 XM_006710944.3 XM_011542248.2 XM_017002474.1 XM_017002475.1 XM_017002476.1 XM_017002477.1 XM_017002478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6364891 c.1198G>A p.E400K Chr.1: 973231-973231 on Build GRCh38
3952727 c.1309-6C>A p.? Chr.1: 973180-973180 on Build GRCh38
6228533 c.1363C>T p.R455C Chr.1: 973240-973240 on Build GRCh38
3952726 c.1153-6C>A p.? Chr.1: 973180-973180 on Build GRCh38
6364892 c.1354G>A p.E452K Chr.1: 973231-973231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557148095 Chr.1: 973100-973100 on Build GRCh38
rs1011368168 Chr.1: 973081-973081 on Build GRCh38
rs76884838 Chr.1: 973205-973205 on Build GRCh38
rs775569859 Chr.1: 973223-973223 on Build GRCh38
rs200388615 Chr.1: 973148-973148 on Build GRCh38
rs1035344145 Chr.1: 973107-973107 on Build GRCh38
rs202212464 Chr.1: 973057-973057 on Build GRCh38
rs559830218 Chr.1: 973201-973201 on Build GRCh38
rs755457620 Chr.1: 973165-973165 on Build GRCh38
rs943938575 Chr.1: 973029-973029 on Build GRCh38
rs767065285 Chr.1: 973137-973137 on Build GRCh38
rs774112267 Chr.1: 973219-973219 on Build GRCh38
rs769741240 Chr.1: 973093-973093 on Build GRCh38
rs760066682 Chr.1: 973147-973147 on Build GRCh38
rs757225927 Chr.1: 973204-973204 on Build GRCh38
rs758993603 Chr.1: 973175-973175 on Build GRCh38
rs768426798 Chr.1: 973056-973056 on Build GRCh38
rs747891825 Chr.1: 973222-973222 on Build GRCh38
rs533063349 Chr.1: 973138-973138 on Build GRCh38
rs777884244 Chr.1: 973022-973022 on Build GRCh38
rs1057488002 Chr.1: 973084-973084 on Build GRCh38
rs747997007 Chr.1: 973044-973044 on Build GRCh38
rs141080484 Chr.1: 973170-973170 on Build GRCh38
rs988892571 Chr.1: 973156-973156 on Build GRCh38
rs866437891 Chr.1: 973192-973192 on Build GRCh38
rs891723864 Chr.1: 973062-973062 on Build GRCh38
rs905611123 Chr.1: 973092-973092 on Build GRCh38
rs375212442 Chr.1: 973207-973207 on Build GRCh38
rs747115705 Chr.1: 973198-973198 on Build GRCh38
rs895456175 Chr.1: 973114-973114 on Build GRCh38
rs1043287168 Chr.1: 973090-973090 on Build GRCh38
rs549085646 Chr.1: 973197-973197 on Build GRCh38
rs201387358 Chr.1: 973144-973144 on Build GRCh38
rs765661942 Chr.1: 973166-973166 on Build GRCh38
rs371927453 Chr.1: 973035-973035 on Build GRCh38
rs771214990 Chr.1: 973028-973028 on Build GRCh38
rs1028271250 Chr.1: 973110-973119 on Build GRCh38
rs28504611 Chr.1: 973034-973034 on Build GRCh38
rs368135287 Chr.1: 973154-973154 on Build GRCh38
rs1019421869 Chr.1: 973145-973145 on Build GRCh38
rs768480355 Chr.1: 973214-973214 on Build GRCh38
rs572032419 Chr.1: 973023-973023 on Build GRCh38
rs886578528 Chr.1: 973085-973085 on Build GRCh38

View Full Product Details