Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 150809359-150809590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHG1
Entrez Gene ID
57480
Gene Name
pleckstrin homology and RhoGEF domain containing G1
Gene Aliases
ARHGEF41
UniGene
Hs.189781
Gene Chromosome Location
Chr.6: 150599863-150843665 on Build GRCh38
Transcripts
NM_001029884.1 XM_005267064.3 XM_005267065.3 XM_005267066.2 XM_006715521.1 XM_006715522.3 XM_011535981.2 XM_011535982.2 XM_011535983.2 XM_017011102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3674578 c.1170C>A p.P390P Chr.6: 150809455-150809455 on Build GRCh38
3019095 c.1191+10G>A p.? Chr.6: 150809486-150809486 on Build GRCh38
4586311 c.1150G>A p.V384I Chr.6: 150809435-150809435 on Build GRCh38
5709906 c.1172A>C p.K391T Chr.6: 150809457-150809457 on Build GRCh38
6287830 c.1150G>T p.V384F Chr.6: 150809435-150809435 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs142544224 Chr.6: 150809446-150809446 on Build GRCh38
rs770199310 Chr.6: 150809557-150809557 on Build GRCh38
rs202011998 Chr.6: 150809431-150809431 on Build GRCh38
rs754408693 Chr.6: 150809501-150809501 on Build GRCh38
rs912802880 Chr.6: 150809566-150809566 on Build GRCh38
rs925406803 Chr.6: 150809534-150809534 on Build GRCh38
rs774591517 Chr.6: 150809460-150809460 on Build GRCh38
rs780934479 Chr.6: 150809515-150809515 on Build GRCh38
rs750161112 Chr.6: 150809517-150809517 on Build GRCh38
rs766856651 Chr.6: 150809494-150809494 on Build GRCh38
rs767812530 Chr.6: 150809484-150809484 on Build GRCh38
rs373235346 Chr.6: 150809520-150809520 on Build GRCh38
rs368681998 Chr.6: 150809510-150809510 on Build GRCh38
rs756948407 Chr.6: 150809486-150809486 on Build GRCh38
rs769584153 Chr.6: 150809433-150809433 on Build GRCh38
rs376176424 Chr.6: 150809502-150809502 on Build GRCh38
rs200133661 Chr.6: 150809422-150809422 on Build GRCh38
rs775628646 Chr.6: 150809435-150809435 on Build GRCh38
rs377233860 Chr.6: 150809525-150809525 on Build GRCh38
rs753981405 Chr.6: 150809483-150809483 on Build GRCh38
rs138892411 Chr.6: 150809445-150809445 on Build GRCh38
rs761893207 Chr.6: 150809478-150809478 on Build GRCh38
rs757946901 Chr.6: 150809485-150809485 on Build GRCh38
rs368515789 Chr.6: 150809556-150809556 on Build GRCh38
rs781492144 Chr.6: 150809520-150809522 on Build GRCh38
rs712216 Chr.6: 150809555-150809555 on Build GRCh38
rs756816892 Chr.6: 150809511-150809511 on Build GRCh38
rs777505892 Chr.6: 150809486-150809486 on Build GRCh38
rs746596797 Chr.6: 150809406-150809406 on Build GRCh38
rs756942947 Chr.6: 150809421-150809421 on Build GRCh38

View Full Product Details