Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45421530-45421763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGT
Entrez Gene ID
51604
Gene Name
phosphatidylinositol glycan anchor biosynthesis class T
Gene Aliases
CGI-06, MCAHS3, NDAP, PNH2
UniGene
Hs.437388
Gene Chromosome Location
Chr.20: 45416067-45426245 on Build GRCh38
Transcripts
NM_001184728.2 NM_001184729.2 NM_001184730.2 NM_015937.5 NR_047691.1 NR_047692.1 NR_047693.1 NR_047694.1 NR_047695.1 XM_005260430.2 XM_005260432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6330880 c.1194C>A p.H398Q Chr.20: 45421543-45421543 on Build GRCh38
5633704 c.1199T>G p.L400R Chr.20: 45421548-45421548 on Build GRCh38
4098835 c.1204A>C p.I402L Chr.20: 45421553-45421553 on Build GRCh38
6261002 c.1224G>C p.E408D Chr.20: 45421573-45421573 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369178179 Chr.20: 45421642-45421642 on Build GRCh38
rs374959599 Chr.20: 45421627-45421627 on Build GRCh38
rs974613670 Chr.20: 45421617-45421617 on Build GRCh38
rs984380032 Chr.20: 45421648-45421648 on Build GRCh38
rs748853753 Chr.20: 45421618-45421618 on Build GRCh38
rs908887897 Chr.20: 45421698-45421698 on Build GRCh38
rs193032432 Chr.20: 45421635-45421635 on Build GRCh38
rs367916073 Chr.20: 45421629-45421629 on Build GRCh38
rs773666314 Chr.20: 45421624-45421624 on Build GRCh38
rs946546009 Chr.20: 45421659-45421659 on Build GRCh38
rs753901894 Chr.20: 45421550-45421550 on Build GRCh38
rs749330243 Chr.20: 45421601-45421601 on Build GRCh38
rs778812534 Chr.20: 45421563-45421563 on Build GRCh38
rs41497050 Chr.20: 45421735-45421735 on Build GRCh38
rs777452786 Chr.20: 45421609-45421609 on Build GRCh38
rs757386694 Chr.20: 45421552-45421552 on Build GRCh38
rs745871702 Chr.20: 45421626-45421626 on Build GRCh38
rs4812899 Chr.20: 45421702-45421702 on Build GRCh38
rs924014746 Chr.20: 45421668-45421668 on Build GRCh38
rs573610505 Chr.20: 45421622-45421622 on Build GRCh38
rs750956280 Chr.20: 45421568-45421568 on Build GRCh38
rs918745171 Chr.20: 45421738-45421738 on Build GRCh38
rs6065815 Chr.20: 45421599-45421599 on Build GRCh38
rs755879674 Chr.20: 45421605-45421605 on Build GRCh38
rs200899089 Chr.20: 45421549-45421549 on Build GRCh38
rs1056515735 Chr.20: 45421677-45421677 on Build GRCh38
rs201261933 Chr.20: 45421619-45421619 on Build GRCh38
rs758913061 Chr.20: 45421573-45421573 on Build GRCh38
rs935405250 Chr.20: 45421676-45421676 on Build GRCh38
rs780635042 Chr.20: 45421591-45421591 on Build GRCh38
rs921797858 Chr.20: 45421644-45421644 on Build GRCh38
rs1044200962 Chr.20: 45421661-45421661 on Build GRCh38
rs559416420 Chr.20: 45421652-45421652 on Build GRCh38
rs867837259 Chr.20: 45421561-45421561 on Build GRCh38
rs577378588 Chr.20: 45421696-45421696 on Build GRCh38

View Full Product Details