Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150194687-150194930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A7
Entrez Gene ID
6534
Gene Name
solute carrier family 6 member 7
Gene Aliases
PROT
UniGene
Hs.241597
Gene Chromosome Location
Chr.5: 150189861-150211072 on Build GRCh38
Transcripts
NM_014228.4 XM_017009767.1 XM_017009768.1 XM_017009769.1 XM_017009770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4727730 c.124G>T p.G42C Chr.5: 150194818-150194818 on Build GRCh38
1672034 c.70C>T p.Q24* Chr.5: 150194764-150194764 on Build GRCh38
4509894 c.81C>T p.V27V Chr.5: 150194775-150194775 on Build GRCh38
7327074 c.84C>T p.D28D Chr.5: 150194778-150194778 on Build GRCh38
6170000 c.217+1G>T p.? Chr.5: 150194912-150194912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765840305 Chr.5: 150194898-150194898 on Build GRCh38
rs866239177 Chr.5: 150194810-150194811 on Build GRCh38
rs754336359 Chr.5: 150194796-150194796 on Build GRCh38
rs751851815 Chr.5: 150194912-150194912 on Build GRCh38
rs764558081 Chr.5: 150194793-150194793 on Build GRCh38
rs914752943 Chr.5: 150194861-150194861 on Build GRCh38
rs146657807 Chr.5: 150194770-150194770 on Build GRCh38
rs377348266 Chr.5: 150194895-150194895 on Build GRCh38
rs545409810 Chr.5: 150194755-150194755 on Build GRCh38
rs774009545 Chr.5: 150194879-150194879 on Build GRCh38
rs766108518 Chr.5: 150194772-150194772 on Build GRCh38
rs202163986 Chr.5: 150194803-150194803 on Build GRCh38
rs777802917 Chr.5: 150194823-150194823 on Build GRCh38
rs906584740 Chr.5: 150194888-150194888 on Build GRCh38
rs201896345 Chr.5: 150194773-150194773 on Build GRCh38
rs377479964 Chr.5: 150194804-150194804 on Build GRCh38
rs139009717 Chr.5: 150194902-150194902 on Build GRCh38
rs746102482 Chr.5: 150194751-150194751 on Build GRCh38
rs763619367 Chr.5: 150194893-150194893 on Build GRCh38
rs1015975879 Chr.5: 150194911-150194911 on Build GRCh38
rs371227337 Chr.5: 150194857-150194857 on Build GRCh38
rs879112115 Chr.5: 150194878-150194878 on Build GRCh38
rs769832668 Chr.5: 150194767-150194767 on Build GRCh38
rs773322357 Chr.5: 150194769-150194769 on Build GRCh38
rs202053910 Chr.5: 150194754-150194754 on Build GRCh38
rs775166670 Chr.5: 150194885-150194885 on Build GRCh38
rs779159357 Chr.5: 150194741-150194741 on Build GRCh38
rs919145365 Chr.5: 150194733-150194733 on Build GRCh38
rs778315192 Chr.5: 150194713-150194713 on Build GRCh38
rs749634463 Chr.5: 150194721-150194721 on Build GRCh38
rs747153518 Chr.5: 150194757-150194757 on Build GRCh38
rs752185966 Chr.5: 150194778-150194778 on Build GRCh38
rs759113293 Chr.5: 150194775-150194775 on Build GRCh38
rs1045549434 Chr.5: 150194846-150194846 on Build GRCh38
rs1004123897 Chr.5: 150194891-150194891 on Build GRCh38
rs192386891 Chr.5: 150194900-150194900 on Build GRCh38
rs771694458 Chr.5: 150194880-150194880 on Build GRCh38
rs199699903 Chr.5: 150194776-150194776 on Build GRCh38
rs747053652 Chr.5: 150194814-150194814 on Build GRCh38
rs749193152 Chr.5: 150194836-150194836 on Build GRCh38
rs780383192 Chr.5: 150194903-150194903 on Build GRCh38
rs374291484 Chr.5: 150194894-150194894 on Build GRCh38
rs757605633 Chr.5: 150194727-150194727 on Build GRCh38
rs760140719 Chr.5: 150194886-150194886 on Build GRCh38
rs768887142 Chr.5: 150194815-150194815 on Build GRCh38
rs755674878 Chr.5: 150194791-150194791 on Build GRCh38
rs757595770 Chr.5: 150194801-150194801 on Build GRCh38
rs780481665 Chr.5: 150194810-150194810 on Build GRCh38

View Full Product Details