Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 127881729-127882002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLK4
Entrez Gene ID
10733
Gene Name
polo like kinase 4
Gene Aliases
MCCRP2, SAK, STK18
UniGene
Hs.172052
Gene Chromosome Location
Chr.4: 127880861-127899224 on Build GRCh38
Transcripts
NM_001190799.1 NM_001190801.1 NM_014264.4 XM_005262701.2 XM_017007662.1 XM_017007663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3010010 c.68T>A p.F23Y Chr.4: 127881868-127881868 on Build GRCh38
7384609 c.74G>T p.G25V Chr.4: 127881874-127881874 on Build GRCh38
5897504 c.61G>A p.G21R Chr.4: 127881861-127881861 on Build GRCh38
7290761 c.118A>G p.I40V Chr.4: 127881918-127881918 on Build GRCh38
4910468 c.55G>T p.G19C Chr.4: 127881855-127881855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769389080 Chr.4: 127881823-127881824 on Build GRCh38
rs756460506 Chr.4: 127881884-127881884 on Build GRCh38
rs369771623 Chr.4: 127881962-127881962 on Build GRCh38
rs773616893 Chr.4: 127881951-127881951 on Build GRCh38
rs765612733 Chr.4: 127881825-127881825 on Build GRCh38
rs144854747 Chr.4: 127881866-127881866 on Build GRCh38
rs552131844 Chr.4: 127881939-127881939 on Build GRCh38
rs745493299 Chr.4: 127881895-127881895 on Build GRCh38
rs761636145 Chr.4: 127881824-127881824 on Build GRCh38
rs769680443 Chr.4: 127881945-127881945 on Build GRCh38
rs778092666 Chr.4: 127881971-127881971 on Build GRCh38
rs769560146 Chr.4: 127881963-127881963 on Build GRCh38
rs776601077 Chr.4: 127881813-127881813 on Build GRCh38
rs771662526 Chr.4: 127881900-127881900 on Build GRCh38
rs541238585 Chr.4: 127881876-127881876 on Build GRCh38
rs758544783 Chr.4: 127881847-127881847 on Build GRCh38
rs751591869 Chr.4: 127881954-127881954 on Build GRCh38
rs377239744 Chr.4: 127881790-127881790 on Build GRCh38
rs185937551 Chr.4: 127881973-127881973 on Build GRCh38
rs781608994 Chr.4: 127881874-127881874 on Build GRCh38
rs201317964 Chr.4: 127881823-127881823 on Build GRCh38
rs772719023 Chr.4: 127881858-127881858 on Build GRCh38
rs766345955 Chr.4: 127881851-127881851 on Build GRCh38
rs1043985991 Chr.4: 127881902-127881902 on Build GRCh38
rs748782005 Chr.4: 127881934-127881938 on Build GRCh38
rs181451128 Chr.4: 127881953-127881953 on Build GRCh38
rs901557104 Chr.4: 127881936-127881936 on Build GRCh38
rs768483997 Chr.4: 127881912-127881912 on Build GRCh38
rs936158959 Chr.4: 127881854-127881854 on Build GRCh38
rs943750506 Chr.4: 127881786-127881786 on Build GRCh38
rs982439572 Chr.4: 127881757-127881757 on Build GRCh38
rs559914338 Chr.4: 127881904-127881904 on Build GRCh38
rs912318797 Chr.4: 127881767-127881767 on Build GRCh38
rs759600174 Chr.4: 127881956-127881956 on Build GRCh38
rs774885670 Chr.4: 127881901-127881901 on Build GRCh38
rs775541700 Chr.4: 127881783-127881783 on Build GRCh38
rs755467547 Chr.4: 127881853-127881853 on Build GRCh38
rs750669030 Chr.4: 127881827-127881827 on Build GRCh38
rs751605196 Chr.4: 127881852-127881852 on Build GRCh38
rs373276627 Chr.4: 127881933-127881933 on Build GRCh38
rs778607661 Chr.4: 127881887-127881887 on Build GRCh38
rs900711609 Chr.4: 127881855-127881855 on Build GRCh38
rs370443589 Chr.4: 127881931-127881931 on Build GRCh38
rs773052247 Chr.4: 127881968-127881968 on Build GRCh38
rs543501670 Chr.4: 127881798-127881798 on Build GRCh38
rs1017451529 Chr.4: 127881755-127881755 on Build GRCh38
rs376790516 Chr.4: 127881803-127881803 on Build GRCh38
rs991968831 Chr.4: 127881764-127881764 on Build GRCh38
rs950026610 Chr.4: 127881775-127881775 on Build GRCh38

View Full Product Details