Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101853351-101853576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFKBIZ
Entrez Gene ID
64332
Gene Name
NFKB inhibitor zeta
Gene Aliases
IKBZ, INAP, MAIL
UniGene
Hs.319171
Gene Chromosome Location
Chr.3: 101827990-101861025 on Build GRCh38
Transcripts
NM_001005474.2 NM_031419.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7244709 c.1015A>G p.S339G Chr.3: 101853541-101853541 on Build GRCh38
6725797 c.994T>C p.S332P Chr.3: 101853520-101853520 on Build GRCh38
6162368 c.1017C>G p.S339R Chr.3: 101853543-101853543 on Build GRCh38
5653474 c.1038T>C p.D346D Chr.3: 101853564-101853564 on Build GRCh38
5397690 c.925G>A p.E309K Chr.3: 101853451-101853451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916823767 Chr.3: 101853382-101853382 on Build GRCh38
rs768773640 Chr.3: 101853389-101853389 on Build GRCh38
rs760057543 Chr.3: 101853492-101853492 on Build GRCh38
rs368611410 Chr.3: 101853486-101853486 on Build GRCh38
rs536323460 Chr.3: 101853392-101853392 on Build GRCh38
rs146210819 Chr.3: 101853377-101853377 on Build GRCh38
rs767317773 Chr.3: 101853542-101853542 on Build GRCh38
rs371473438 Chr.3: 101853548-101853548 on Build GRCh38
rs761048352 Chr.3: 101853432-101853432 on Build GRCh38
rs1000263983 Chr.3: 101853551-101853551 on Build GRCh38
rs35313454 Chr.3: 101853384-101853384 on Build GRCh38
rs142122122 Chr.3: 101853399-101853399 on Build GRCh38
rs184488416 Chr.3: 101853440-101853440 on Build GRCh38
rs925414417 Chr.3: 101853423-101853423 on Build GRCh38
rs779015430 Chr.3: 101853475-101853475 on Build GRCh38
rs775094689 Chr.3: 101853502-101853502 on Build GRCh38
rs777684350 Chr.3: 101853376-101853376 on Build GRCh38
rs116659590 Chr.3: 101853504-101853504 on Build GRCh38
rs148590449 Chr.3: 101853487-101853487 on Build GRCh38
rs138046725 Chr.3: 101853521-101853521 on Build GRCh38
rs374457523 Chr.3: 101853456-101853456 on Build GRCh38
rs772320025 Chr.3: 101853420-101853420 on Build GRCh38
rs774349082 Chr.3: 101853395-101853395 on Build GRCh38
rs374702097 Chr.3: 101853552-101853552 on Build GRCh38
rs199980845 Chr.3: 101853466-101853466 on Build GRCh38
rs368288845 Chr.3: 101853435-101853435 on Build GRCh38
rs3821727 Chr.3: 101853446-101853446 on Build GRCh38
rs146355720 Chr.3: 101853473-101853473 on Build GRCh38
rs763865646 Chr.3: 101853460-101853460 on Build GRCh38
rs749258201 Chr.3: 101853386-101853386 on Build GRCh38
rs766736566 Chr.3: 101853438-101853438 on Build GRCh38
rs757025168 Chr.3: 101853526-101853526 on Build GRCh38
rs752189133 Chr.3: 101853453-101853453 on Build GRCh38
rs748344072 Chr.3: 101853414-101853414 on Build GRCh38
rs745801101 Chr.3: 101853468-101853468 on Build GRCh38
rs765693919 Chr.3: 101853498-101853498 on Build GRCh38
rs780913282 Chr.3: 101853467-101853467 on Build GRCh38
rs199648787 Chr.3: 101853434-101853434 on Build GRCh38
rs763775403 Chr.3: 101853506-101853506 on Build GRCh38
rs202168485 Chr.3: 101853483-101853483 on Build GRCh38
rs764288627 Chr.3: 101853478-101853478 on Build GRCh38
rs773434450 Chr.3: 101853485-101853485 on Build GRCh38
rs773524436 Chr.3: 101853425-101853425 on Build GRCh38

View Full Product Details