Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207861054-207861325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHM3
Entrez Gene ID
389072
Gene Name
pleckstrin homology domain containing M3
Gene Aliases
DAPR, PLEKHM1L
UniGene
Hs.159188
Gene Chromosome Location
Chr.2: 207821288-208025675 on Build GRCh38
Transcripts
NM_001080475.2 XM_005246545.3 XM_011511160.2 XM_017004072.1 XM_017004073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6410574 c.2016C>A p.Y672* Chr.2: 207861197-207861197 on Build GRCh38
3576918 c.1990A>G p.I664V Chr.2: 207861223-207861223 on Build GRCh38
3991007 c.2033G>C p.S678T Chr.2: 207861180-207861180 on Build GRCh38
5109341 c.1979T>C p.L660S Chr.2: 207861234-207861234 on Build GRCh38
719554 c.1960G>T p.G654* Chr.2: 207861253-207861253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779514675 Chr.2: 207861293-207861293 on Build GRCh38
rs760295595 Chr.2: 207861252-207861252 on Build GRCh38
rs758550099 Chr.2: 207861280-207861280 on Build GRCh38
rs752091040 Chr.2: 207861275-207861275 on Build GRCh38
rs754349912 Chr.2: 207861097-207861097 on Build GRCh38
rs778046594 Chr.2: 207861130-207861130 on Build GRCh38
rs764122343 Chr.2: 207861283-207861283 on Build GRCh38
rs747265280 Chr.2: 207861146-207861146 on Build GRCh38
rs768607642 Chr.2: 207861202-207861202 on Build GRCh38
rs765856399 Chr.2: 207861278-207861278 on Build GRCh38
rs765368305 Chr.2: 207861112-207861112 on Build GRCh38
rs766718739 Chr.2: 207861272-207861272 on Build GRCh38
rs758640638 Chr.2: 207861122-207861122 on Build GRCh38
rs1034739436 Chr.2: 207861197-207861197 on Build GRCh38
rs370843765 Chr.2: 207861199-207861199 on Build GRCh38
rs910490228 Chr.2: 207861176-207861176 on Build GRCh38
rs755909085 Chr.2: 207861157-207861157 on Build GRCh38
rs200896303 Chr.2: 207861116-207861116 on Build GRCh38
rs771626973 Chr.2: 207861231-207861231 on Build GRCh38
rs753318137 Chr.2: 207861279-207861279 on Build GRCh38
rs755565274 Chr.2: 207861099-207861099 on Build GRCh38
rs185632032 Chr.2: 207861203-207861203 on Build GRCh38
rs772911934 Chr.2: 207861239-207861239 on Build GRCh38
rs990909634 Chr.2: 207861244-207861244 on Build GRCh38
rs762422346 Chr.2: 207861255-207861255 on Build GRCh38
rs761085602 Chr.2: 207861088-207861088 on Build GRCh38
rs755791889 Chr.2: 207861287-207861287 on Build GRCh38
rs773390346 Chr.2: 207861084-207861084 on Build GRCh38
rs199543358 Chr.2: 207861095-207861095 on Build GRCh38
rs375277146 Chr.2: 207861200-207861200 on Build GRCh38
rs747669781 Chr.2: 207861224-207861224 on Build GRCh38
rs776892534 Chr.2: 207861276-207861276 on Build GRCh38

View Full Product Details