Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8522562-8522831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO1F
Entrez Gene ID
4542
Gene Name
myosin IF
Gene Aliases
-
UniGene
Hs.465818
Gene Chromosome Location
Chr.19: 8520790-8577447 on Build GRCh38
Transcripts
NM_012335.3 XM_011528024.1 XM_011528025.1 XM_011528026.1 XM_011528027.1 XM_017026821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5441080 c.3040G>A p.G1014S Chr.19: 8522644-8522644 on Build GRCh38
4624176 c.2993G>A p.R998H Chr.19: 8522691-8522691 on Build GRCh38
5674308 c.2919T>A p.S973S Chr.19: 8522765-8522765 on Build GRCh38
5678188 c.2885C>T p.A962V Chr.19: 8522799-8522799 on Build GRCh38
7378512 c.2983C>T p.P995S Chr.19: 8522701-8522701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549007882 Chr.19: 8522662-8522662 on Build GRCh38
rs769512460 Chr.19: 8522759-8522759 on Build GRCh38
rs555693888 Chr.19: 8522790-8522790 on Build GRCh38
rs762996920 Chr.19: 8522696-8522696 on Build GRCh38
rs750731175 Chr.19: 8522733-8522733 on Build GRCh38
rs942932058 Chr.19: 8522734-8522734 on Build GRCh38
rs758965119 Chr.19: 8522735-8522735 on Build GRCh38
rs570160376 Chr.19: 8522730-8522730 on Build GRCh38
rs549991223 Chr.19: 8522722-8522722 on Build GRCh38
rs768230288 Chr.19: 8522683-8522683 on Build GRCh38
rs767709488 Chr.19: 8522624-8522624 on Build GRCh38
rs776353331 Chr.19: 8522612-8522612 on Build GRCh38
rs754535521 Chr.19: 8522740-8522740 on Build GRCh38
rs374018985 Chr.19: 8522635-8522635 on Build GRCh38
rs770815982 Chr.19: 8522785-8522785 on Build GRCh38
rs757596507 Chr.19: 8522639-8522639 on Build GRCh38
rs769881987 Chr.19: 8522812-8522812 on Build GRCh38
rs747807146 Chr.19: 8522743-8522743 on Build GRCh38
rs201056652 Chr.19: 8522629-8522629 on Build GRCh38
rs775501333 Chr.19: 8522698-8522698 on Build GRCh38
rs779200718 Chr.19: 8522654-8522654 on Build GRCh38
rs200159025 Chr.19: 8522772-8522772 on Build GRCh38
rs1033645328 Chr.19: 8522613-8522613 on Build GRCh38
rs754112921 Chr.19: 8522723-8522723 on Build GRCh38
rs377201964 Chr.19: 8522673-8522673 on Build GRCh38
rs967230317 Chr.19: 8522788-8522788 on Build GRCh38
rs368435303 Chr.19: 8522636-8522636 on Build GRCh38
rs773197400 Chr.19: 8522762-8522762 on Build GRCh38
rs765311016 Chr.19: 8522622-8522622 on Build GRCh38
rs376499204 Chr.19: 8522586-8522586 on Build GRCh38
rs1025207126 Chr.19: 8522594-8522594 on Build GRCh38
rs776630100 Chr.19: 8522809-8522809 on Build GRCh38
rs780576676 Chr.19: 8522739-8522739 on Build GRCh38
rs201578688 Chr.19: 8522599-8522599 on Build GRCh38
rs370097600 Chr.19: 8522588-8522588 on Build GRCh38
rs547685891 Chr.19: 8522687-8522687 on Build GRCh38
rs370594416 Chr.19: 8522625-8522625 on Build GRCh38
rs975908245 Chr.19: 8522756-8522788 on Build GRCh38
rs893819278 Chr.19: 8522756-8522756 on Build GRCh38
rs367779407 Chr.19: 8522620-8522620 on Build GRCh38
rs759344792 Chr.19: 8522791-8522791 on Build GRCh38
rs73922133 Chr.19: 8522669-8522669 on Build GRCh38
rs1041231591 Chr.19: 8522737-8522737 on Build GRCh38
rs910231638 Chr.19: 8522679-8522679 on Build GRCh38
rs2288411 Chr.19: 8522805-8522805 on Build GRCh38
rs552336969 Chr.19: 8522657-8522657 on Build GRCh38
rs764273527 Chr.19: 8522703-8522703 on Build GRCh38
rs773074453 Chr.19: 8522692-8522692 on Build GRCh38
rs199971487 Chr.19: 8522691-8522691 on Build GRCh38
rs376454294 Chr.19: 8522697-8522697 on Build GRCh38
rs997215193 Chr.19: 8522603-8522603 on Build GRCh38
rs201007272 Chr.19: 8522729-8522729 on Build GRCh38
rs368617557 Chr.19: 8522688-8522688 on Build GRCh38
rs750405338 Chr.19: 8522623-8522623 on Build GRCh38

View Full Product Details