Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23371691-23371964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCNN1B
Entrez Gene ID
6338
Gene Name
sodium channel epithelial 1 beta subunit
Gene Aliases
BESC1, ENaCb, ENaCbeta, SCNEB
UniGene
Hs.414614
Gene Chromosome Location
Chr.16: 23278231-23381299 on Build GRCh38
Transcripts
NM_000336.2 XM_011545913.2 XM_011545914.1 XM_017023525.1 XM_017023526.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4520523 c.1197G>A p.M399I Chr.16: 23371793-23371793 on Build GRCh38
1646748 c.1188T>C p.L396L Chr.16: 23371784-23371784 on Build GRCh38
1678932 c.1193G>A p.R398H Chr.16: 23371789-23371789 on Build GRCh38
3957341 c.1121A>T p.Y374F Chr.16: 23371852-23371852 on Build GRCh38
3957342 c.1256A>T p.Y419F Chr.16: 23371852-23371852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72654342 Chr.16: 23371912-23371912 on Build GRCh38
rs755232411 Chr.16: 23371873-23371873 on Build GRCh38
rs137852711 Chr.16: 23371836-23371836 on Build GRCh38
rs867187844 Chr.16: 23371877-23371877 on Build GRCh38
rs150949875 Chr.16: 23371788-23371788 on Build GRCh38
rs774130659 Chr.16: 23371817-23371817 on Build GRCh38
rs370595048 Chr.16: 23371900-23371900 on Build GRCh38
rs762142010 Chr.16: 23371888-23371889 on Build GRCh38
rs41278184 Chr.16: 23371867-23371867 on Build GRCh38
rs1012894741 Chr.16: 23371920-23371920 on Build GRCh38
rs781738224 Chr.16: 23371884-23371884 on Build GRCh38
rs994871969 Chr.16: 23371902-23371902 on Build GRCh38
rs755499367 Chr.16: 23371789-23371789 on Build GRCh38
rs932297365 Chr.16: 23371800-23371800 on Build GRCh38
rs144780745 Chr.16: 23371843-23371843 on Build GRCh38
rs750710383 Chr.16: 23371841-23371841 on Build GRCh38
rs200719352 Chr.16: 23371892-23371892 on Build GRCh38
rs765382334 Chr.16: 23371839-23371839 on Build GRCh38
rs761731857 Chr.16: 23371746-23371746 on Build GRCh38
rs758879922 Chr.16: 23371765-23371765 on Build GRCh38
rs1028479510 Chr.16: 23371724-23371724 on Build GRCh38
rs769150286 Chr.16: 23371804-23371804 on Build GRCh38
rs890904629 Chr.16: 23371749-23371749 on Build GRCh38
rs778468492 Chr.16: 23371914-23371914 on Build GRCh38
rs748918495 Chr.16: 23371811-23371811 on Build GRCh38
rs747701524 Chr.16: 23371791-23371791 on Build GRCh38
rs765587348 Chr.16: 23371753-23371753 on Build GRCh38
rs370750866 Chr.16: 23371913-23371913 on Build GRCh38
rs373477307 Chr.16: 23371761-23371761 on Build GRCh38
rs72654341 Chr.16: 23371893-23371893 on Build GRCh38
rs72654343 Chr.16: 23371925-23371925 on Build GRCh38
rs777426154 Chr.16: 23371810-23371810 on Build GRCh38
rs770754286 Chr.16: 23371815-23371815 on Build GRCh38
rs761974708 Chr.16: 23371838-23371838 on Build GRCh38
rs2303157 Chr.16: 23371926-23371926 on Build GRCh38
rs775547565 Chr.16: 23371933-23371933 on Build GRCh38
rs142550343 Chr.16: 23371818-23371818 on Build GRCh38
rs917991214 Chr.16: 23371880-23371880 on Build GRCh38
rs1129412 Chr.16: 23371778-23371778 on Build GRCh38
rs1036469535 Chr.16: 23371863-23371863 on Build GRCh38
rs1015247952 Chr.16: 23371780-23371780 on Build GRCh38
rs372993301 Chr.16: 23371871-23371871 on Build GRCh38
rs777702262 Chr.16: 23371922-23371922 on Build GRCh38
rs376646015 Chr.16: 23371770-23371770 on Build GRCh38
rs751969143 Chr.16: 23371868-23371868 on Build GRCh38
rs772050858 Chr.16: 23371825-23371825 on Build GRCh38
rs1054242678 Chr.16: 23371773-23371773 on Build GRCh38
rs924465445 Chr.16: 23371717-23371717 on Build GRCh38

View Full Product Details